Search Thermo Fisher Scientific
Immunogen sequence: MTEHRRDEE NARLLAELVR ARGLKLDGCF SYWDDCLVLT ALLCQELGLP CSSPAALRLA KQKSLTQLHL LHHHGPPWPA PSLHAVPCCP LESEADVERA VHQVPLPGVM KLEFGAGAVG VRLVEDAPQC HEHFSRITRD LQGEADHPGI GLGWGNAMLL MEFVEGTEHD VDLVLFGGRL LAAFVSDNGP TRLPGFTETA ACMPTGLAPE QEAQMVQAAF RCCLGCGLLD GVFNVELKLT GAGPRLIEIN PRMGGFYLRD WILELYGVDL LLAAVMVACG LRPALPTRPR ARGHLVGVMC LVSQHLQALS STASRETLQA LHDRGLLRLN LLEEALVPGE YEEPYCSVAC AGPSPTEARL RLLGLCQGLG IDGPSYPVAH FLSHFK (443-827 aa encoded by BC036557)
CARNS1 (EC 6.3.2.11), a member of the ATP-grasp family of ATPases, catalyzes the formation of carnosine (beta-alanyl-L-histidine) and homocarnosine (gamma-aminobutyryl-L-histidine), which are found mainly in skeletal muscle and the central nervous system, respectively.