Product References
Engineered bone marrow as a clinically relevant ex vivo model for primary bone cancer research and drug screening.
Proceedings of the National Academy of Sciences of the United States of America
Griffin KH,Thorpe SW,Sebastian A,Hum NR,Coonan TP,Sagheb IS,Loots GG,Randall RL,Leach JK
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Sep 26 00:00:00 EDT 2023
Laxative effect of Wenyang Yiqi Decoction on loperamide-induced astriction model mice.
Annals of translational medicine
Su Y,Zhu R,Pang C,He Z,Wu B,Wang X
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Western Blot
Tue Feb 28 00:00:00 EST 2023
The AML-associated K313 mutation enhances C/EBPα activity by leading to C/EBPα overexpression.
Cell death & disease
Gentle IE,Moelter I,Badr MT,Döhner K,Lübbert M,Häcker G
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
The lifespan quantitative trait locus gene Securin controls hematopoietic progenitor cell function.
Haematologica
Brown A,Schuetz D,Han Y,Daria D,Nattamai KJ,Eiwen K,Sakk V,Pospiech J,Saller T,van Zant G,Wagner W,Geiger H
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Apr 27 00:00:00 EDT 2021
Morphological and immunohistochemical phenotype of TCs in the intestinal bulb of Grass carp and their potential role in intestinal immunity.
Scientific reports
Abd-Elhafeez HH,Abou-Elhamd AS,Soliman SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Thu Aug 20 00:00:00 EDT 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Inhibition of FGF2-Mediated Signaling in GIST-Promising Approach for Overcoming Resistance to Imatinib.
Cancers
Boichuk S,Galembikova A,Mikheeva E,Bikinieva F,Aukhadieva A,Dunaev P,Khalikov D,Petrov S,Kurtasanov R,Valeeva E,Kireev I,Dugina V,Lushnikova A,Novikova M,Kopnin P
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Wed Jun 24 00:00:00 EDT 2020
Fish telocytes and their relation to rodlet cells in ruby-red-fin shark (rainbow shark) Epalzeorhynchos frenatum (Teleostei: Cyprinidae).
Scientific reports
Abd-Elhafeez HH,Abdo W,Kamal BM,Soliman SA
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Tue Nov 03 00:00:00 EST 2020
Loss of FGFR3 Delays Acute Myeloid Leukemogenesis by Programming Weakly Pathogenic CD117-Positive Leukemia Stem-Like Cells.
Frontiers in pharmacology
Guo C,Ran Q,Sun C,Zhou T,Yang X,Zhang J,Pang S,Xiao Y
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Feb 16 00:00:00 EST 2021
Hsd3b2 associated in modulating steroid hormone synthesis pathway regulates the differentiation of chicken embryonic stem cells into spermatogonial stem cells.
Journal of cellular biochemistry
Zhang C,Wang M,He N,Ahmed MF,Wang Y,Zhao R,Yu X,Jin J,Song J,Zuo Q,Zhang Y,Li B
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunocytochemistry
Mon Jan 01 00:00:00 EST 2018
Electroacupuncture at ST36 Increases Bone Marrow-Derived Interstitial Cells of Cajal via the SDF-1/CXCR4 and mSCF/Kit-ETV1 Pathways in the Stomach of Diabetic Mice.
Evidence-based complementary and alternative medicine : eCAM
Zhao J,An J,Liu S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Wed Mar 16 00:00:00 EDT 2022
Bioengineering functional smooth muscle with spontaneous rhythmic contraction in vitro.
Scientific reports
Kobayashi M,Khalil HA,Lei NY,Wang Q,Wang K,Wu BM,Dunn JCY
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Mon Sep 10 00:00:00 EDT 2018
Bioengineering functional smooth muscle with spontaneous rhythmic contraction in vitro.
Scientific reports
Kobayashi M,Khalil HA,Lei NY,Wang Q,Wang K,Wu BM,Dunn JCY
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Mon Sep 10 00:00:00 EDT 2018
c-Kit+ Cells in Adult Salivary Glands do not Function as Tissue Stem Cells.
Scientific reports
Kwak M,Ninche N,Klein S,Saur D,Ghazizadeh S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Fri Sep 21 00:00:00 EDT 2018
GATA4-dependent organ-specific endothelial differentiation controls liver development and embryonic hematopoiesis.
The Journal of clinical investigation
Géraud C,Koch PS,Zierow J,Klapproth K,Busch K,Olsavszky V,Leibing T,Demory A,Ulbrich F,Diett M,Singh S,Sticht C,Breitkopf-Heinlein K,Richter K,Karppinen SM,Pihlajaniemi T,Arnold B,Rodewald HR,Augustin HG,Schledzewski K,Goerdt S
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2017
Conditional genetic deletion of Ano1 in interstitial cells of Cajal impairs Ca2+ transients and slow waves in adult mouse small intestine.
American journal of physiology. Gastrointestinal and liver physiology
Malysz J,Gibbons SJ,Saravanaperumal SA,Du P,Eisenman ST,Cao C,Oh U,Saur D,Klein S,Ordog T,Farrugia G
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Wed Mar 01 00:00:00 EST 2017
Low Testosterone Alters the Activity of Mouse Prostate Stem Cells.
The Prostate
Zhou Y,Copeland B,Otto-Duessel M,He M,Markel S,Synold TW,Jones JO
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
A Functional Bioluminescent Zebrafish Screen for Enhancing Hematopoietic Cell Homing.
Stem cell reports
Astuti Y,Kramer AC,Blake AL,Blazar BR,Tolar J,Taisto ME,Lund TC
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Jan 10 00:00:00 EST 2017
Mutant U2AF1-expressing cells are sensitive to pharmacological modulation of the spliceosome.
Nature communications
Shirai CL,White BS,Tripathi M,Tapia R,Ley JN,Ndonwi M,Kim S,Shao J,Carver A,Saez B,Fulton RS,Fronick C,O'Laughlin M,Lagisetti C,Webb TR,Graubert TA,Walter MJ
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Mon Jan 09 00:00:00 EST 2017
Core Circadian Clock Genes Regulate Leukemia Stem Cells in AML.
Cell
Puram RV,Kowalczyk MS,de Boer CG,Schneider RK,Miller PG,McConkey M,Tothova Z,Tejero H,Heckl D,Järås M,Chen MC,Li H,Tamayo A,Cowley GS,Rozenblatt-Rosen O,Al-Shahrour F,Regev A,Ebert BL
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Thu Apr 07 00:00:00 EDT 2016
Stimulus-induced pacemaker activity in interstitial cells of Cajal associated with the deep muscular plexus of the small intestine.
Neurogastroenterology and motility
Zhu YF,Wang XY,Parsons SP,Huizinga JD
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunocytochemistry
Fri Jul 01 00:00:00 EDT 2016
Antagonism of stem cell factor/c-kit signaling attenuates neonatal chronic hypoxia-induced pulmonary vascular remodeling.
Pediatric research
Young KC,Torres E,Hehre D,Wu S,Suguihara C,Hare JM
16-1172 was used in Neutralization experiments to study how SCF/c-kit signaling may potentiate chronic hypoxia-induced vascular remodeling by modulating ERK activation.
Fri Apr 01 00:00:00 EDT 2016
Regulation of B-cell development and tolerance by different members of the miR-17∼92 family microRNAs.
Nature communications
Lai M,Gonzalez-Martin A,Cooper AB,Oda H,Jin HY,Shepherd J,He L,Zhu J,Nemazee D,Xiao C
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Aug 02 00:00:00 EDT 2016
Transcription factor c-Rel plays a crucial role in driving anti-CD40-mediated innate colitis.
Mucosal immunology
Visekruna A,Linnerz T,Martinic V,Vachharajani N,Hartmann S,Harb H,Joeris T,Pfefferle PI,Hofer MJ,Steinhoff U
16-1172 was used in Flow cytometry/Cell sorting to investigate the role c-Rel plays in regulating innate lymphoid cells which induce anti-CD40-mediated innate colitis in mice.
Sun Mar 01 00:00:00 EST 2015
The origin of segmentation motor activity in the intestine.
Nature communications
Huizinga JD,Chen JH,Zhu YF,Pawelka A,McGinn RJ,Bardakjian BL,Parsons SP,Kunze WA,Wu RY,Bercik P,Khoshdel A,Chen S,Yin S,Zhang Q,Yu Y,Gao Q,Li K,Hu X,Zarate N,Collins P,Pistilli M,Ma J,Zhang R,Chen D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Mon Nov 02 00:00:00 EST 2015
Selective regulation of lymphopoiesis and leukemogenesis by individual zinc fingers of Ikaros.
Nature immunology
Schjerven H,McLaughlin J,Arenzana TL,Frietze S,Cheng D,Wadsworth SE,Lawson GW,Bensinger SJ,Farnham PJ,Witte ON,Smale ST
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
TRPA1-dependent pruritus in IL-13-induced chronic atopic dermatitis.
Journal of immunology (Baltimore, Md. : 1950)
Oh MH,Oh SY,Lu J,Lou H,Myers AC,Zhu Z,Zheng T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Sun Dec 01 00:00:00 EST 2013
TRPA1-dependent pruritus in IL-13-induced chronic atopic dermatitis.
Journal of immunology (Baltimore, Md. : 1950)
Oh MH,Oh SY,Lu J,Lou H,Myers AC,Zhu Z,Zheng T
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Sun Dec 01 00:00:00 EST 2013
Cells derived from the coelomic epithelium contribute to multiple gastrointestinal tissues in mouse embryos.
PloS one
Carmona R,Cano E,Mattiotti A,Gaztambide J,Muñoz-Chápuli R
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Fri Aug 09 00:00:00 EDT 2013
Mast cell deficiency attenuates progression of atherosclerosis and hepatic steatosis in apolipoprotein E-null mice.
American journal of physiology. Heart and circulatory physiology
Smith DD,Tan X,Raveendran VV,Tawfik O,Stechschulte DJ,Dileepan KN
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunohistochemistry
Fri Jun 15 00:00:00 EDT 2012
Homeostatic role of transforming growth factor-beta in the oral cavity and esophagus of mice and its expression by mast cells in these tissues.
The American journal of pathology
Vitsky A,Waire J,Pawliuk R,Bond A,Matthews D,Lacasse E,Hawes ML,Nelson C,Richards S,Piepenhagen PA,Garman RD,Andrews L,Thurberg BL,Lonning S,Ledbetter S,Ruzek MC
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Immunofluorescence
Mon Jun 01 00:00:00 EDT 2009
Efficient transplantation via antibody-based clearance of hematopoietic stem cell niches.
Science (New York, N.Y.)
Czechowicz A,Kraft D,Weissman IL,Bhattacharya D
Published figure using CD117 (c-Kit) monoclonal antibody (Product # 16-1172-82) in Flow Cytometry
Fri Nov 23 00:00:00 EST 2007