Product References
PI3Kδ coordinates transcriptional, chromatin, and metabolic changes to promote effector CD8+ T cells at the expense of central memory.
Cell reports
Cannons JL,Villarino AV,Kapnick SM,Preite S,Shih HY,Gomez-Rodriguez J,Kaul Z,Shibata H,Reilley JM,Huang B,Handon R,McBain IT,Gossa S,Wu T,Su HC,McGavern DB,O'Shea JJ,McGuire PJ,Uzel G,Schwartzberg PL
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Tue Oct 12 00:00:00 EDT 2021
Hedgehog interacting protein-expressing lung fibroblasts suppress lymphocytic inflammation in mice.
JCI insight
Yun JH,Lee C,Liu T,Liu S,Kim EY,Xu S,Curtis JL,Pinello L,Bowler RP,Silverman EK,Hersh CP,Zhou X
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Wed Sep 08 00:00:00 EDT 2021
RIPK3 and Caspase-1/11 Are Necessary for Optimal Antigen-Specific CD8 T Cell Response Elicited by Genetically Modified Listeria monocytogenes.
Frontiers in immunology
Rana A,de Almeida FC,Paico Montero HA,Gonzales Carazas MM,Bortoluci KR,Sad S,Amarante-Mendes GP
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Fri Mar 26 00:00:00 EDT 2021
RNA Demethylase ALKBH5 Selectively Promotes Tumorigenesis and Cancer Stem Cell Self-Renewal in Acute Myeloid Leukemia.
Cell stem cell
Shen C,Sheng Y,Zhu AC,Robinson S,Jiang X,Dong L,Chen H,Su R,Yin Z,Li W,Deng X,Chen Y,Hu YC,Weng H,Huang H,Prince E,Cogle CR,Sun M,Zhang B,Chen CW,Marcucci G,He C,Qian Z,Chen J
13-1271-82 was used in Flow Cytometry to reveal crucial functions of ALKBH5 in leukemogenesis and LSC/LIC self-renewal/maintenance and highlight the therapeutic potential of targeting the ALKBH5/m6A axis.
Thu Jul 02 00:00:00 EDT 2020
Long-term ex vivo expansion of mouse hematopoietic stem cells.
Nature protocols
Wilkinson AC,Ishida R,Nakauchi H,Yamazaki S
13-1271 was used in Flow cytometry/Cell sorting to describe a simple platform for the expansion of functional mouse hematopoietic stem cells ex vivo for >1 month under fully defined albumin-free conditions.
Sat Feb 01 00:00:00 EST 2020
Single-Cell Profiling Defines Transcriptomic Signatures Specific to Tumor-Reactive versus Virus-Responsive CD4+ T Cells.
Cell reports
Magen A,Nie J,Ciucci T,Tamoutounour S,Zhao Y,Mehta M,Tran B,McGavern DB,Hannenhalli S,Bosselut R
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Tue Dec 03 00:00:00 EST 2019
Bone marrow adipose tissue-derived stem cell factor mediates metabolic regulation of hematopoiesis.
Haematologica
Zhang Z,Huang Z,Ong B,Sahu C,Zeng H,Ruan HB
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Long-term ex vivo haematopoietic-stem-cell expansion allows nonconditioned transplantation.
Nature
Wilkinson AC,Ishida R,Kikuchi M,Sudo K,Morita M,Crisostomo RV,Yamamoto R,Loh KM,Nakamura Y,Watanabe M,Nakauchi H,Yamazaki S
13-1271 was used in Flow cytometry/Cell sorting to identify polyvinyl alcohol as a functionally superior replacement for serum albumin that is compatible with good manufacturing practice.
Mon Jul 01 00:00:00 EDT 2019
An Id2RFP-Reporter Mouse Redefines Innate Lymphoid Cell Precursor Potentials.
Immunity
Xu W,Cherrier DE,Chea S,Vosshenrich C,Serafini N,Petit M,Liu P,Golub R,Di Santo JP
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Tue Apr 16 00:00:00 EDT 2019
Metabolic Imaging Reveals a Unique Preference of Symmetric Cell Division and Homing of Leukemia-Initiating Cells in an Endosteal Niche.
Cell metabolism
Hao X,Gu H,Chen C,Huang D,Zhao Y,Xie L,Zou Y,Shu HS,Zhang Y,He X,Lai X,Zhang X,Zhou BO,Zhang CC,Chen GQ,Yu Z,Yang Y,Zheng J
13-1271 was used in Flow cytometry/Cell sorting to provide a unique angle for the study of metabolisms in stem cells, and may lead to development of novel strategies for cancer treatment.
Tue Apr 02 00:00:00 EDT 2019
Activation of naïve CD4+ T cells re-tunes STAT1 signaling to deliver unique cytokine responses in memory CD4+ T cells.
Nature immunology
Twohig JP,Cardus Figueras A,Andrews R,Wiede F,Cossins BC,Derrac Soria A,Lewis MJ,Townsend MJ,Millrine D,Li J,Hill DG,Uceda Fernandez J,Liu X,Szomolay B,Pepper CJ,Taylor PR,Pitzalis C,Tiganis T,Williams NM,Jones GW,Jones SA
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2019
The interplay of BMP4 and IL‑7 regulates the apoptosis of intestinal intraepithelial lymphocytes under conditions of ischemia̸reperfusion.
International journal of molecular medicine
Luo B,Chen K,Feng Q,Xiao W,Ma D,Yang H,Zhang C
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Tue May 01 00:00:00 EDT 2018
Large-Scale Clonal Analysis Resolves Aging of the Mouse Hematopoietic Stem Cell Compartment.
Cell stem cell
Yamamoto R,Wilkinson AC,Ooehara J,Lan X,Lai CY,Nakauchi Y,Pritchard JK,Nakauchi H
13-1271 was used in Flow cytometry/Cell sorting to determine the age-related functional changes to the haematopoietic stem cell compartment.
Thu Apr 05 00:00:00 EDT 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Oncofetal gene SALL4 reactivation by hepatitis B virus counteracts miR-200c in PD-L1-induced T cell exhaustion.
Nature communications
Sun C,Lan P,Han Q,Huang M,Zhang Z,Xu G,Song J,Wang J,Wei H,Zhang J,Sun R,Zhang C,Tian Z
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Wed Mar 28 00:00:00 EDT 2018
Regulatory Innate Lymphoid Cells Control Innate Intestinal Inflammation.
Cell
Wang S,Xia P,Chen Y,Qu Y,Xiong Z,Ye B,Du Y,Tian Y,Yin Z,Xu Z,Fan Z
13-1271 was used in Immunohistochemistry-immunofluorescence to identify a regulatory subpopulation of innate lymphoid cells that exists in the gut, and harbour a unique gene identity.
Thu Sep 21 00:00:00 EDT 2017
SWEF Proteins Distinctly Control Maintenance and Differentiation of Hematopoietic Stem Cells.
PloS one
Ripich T,Chacón-Martínez CA,Fischer L,Pernis A,Kiessling N,Garbe AI,Jessberger R
13-1271 was used in Flow cytometry/Cell sorting to investigate the role of SWAP-70 and DEF6 in hematopoietic stem and precursor cells.
Mon Jul 31 00:00:00 EDT 2017
Identification of Nascent Memory CD8 T Cells and Modeling of Their Ontogeny.
Cell systems
Crauste F,Mafille J,Boucinha L,Djebali S,Gandrillon O,Marvel J,Arpin C
13-1271 was used in Flow cytometry/Cell sorting to identify Bcl2 and Mki67 as a marker combination that enables the tracking of nascent memory cells within the effector phase.
Wed Mar 22 00:00:00 EDT 2017
A simple, accurate and universal method for quantification of PCR.
BMC biotechnology
Boulter N,Suarez FG,Schibeci S,Sunderland T,Tolhurst O,Hunter T,Hodge G,Handelsman D,Simanainen U,Hendriks E,Duggan K
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Wed Mar 09 00:00:00 EST 2016
Production of IL-10 by CD4(+) regulatory T cells during the resolution of infection promotes the maturation of memory CD8(+) T cells.
Nature immunology
Laidlaw BJ,Cui W,Amezquita RA,Gray SM,Guan T,Lu Y,Kobayashi Y,Flavell RA,Kleinstein SH,Craft J,Kaech SM
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2015
T Cell factor 1 represses CD8+ effector T cell formation and function.
Journal of immunology (Baltimore, Md. : 1950)
Tiemessen MM,Baert MR,Kok L,van Eggermond MC,van den Elsen PJ,Arens R,Staal FJ
13-1271 was used in Flow cytometry/Cell sorting to explore the role of the Wnt-responsive transcription factor T cell factor 1 in the initial phases of CD8(+) T effector cell formation.
Mon Dec 01 00:00:00 EST 2014
WASH is required for the differentiation commitment of hematopoietic stem cells in a c-Myc-dependent manner.
The Journal of experimental medicine
Xia P,Wang S,Huang G,Zhu P,Li M,Ye B,Du Y,Fan Z
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Mon Sep 22 00:00:00 EDT 2014
Highly self-reactive naive CD4 T cells are prone to differentiate into regulatory T cells.
Nature communications
Martin B,Auffray C,Delpoux A,Pommier A,Durand A,Charvet C,Yakonowsky P,de Boysson H,Bonilla N,Audemard A,Sparwasser T,Salomon BL,Malissen B,Lucas B
13-1271 was used in Flow cytometry/Cell sorting to show that naive CD4 T cells with the highest avidity for self are prone to differentiate into regulatory T cells.
Thu Feb 27 00:00:00 EST 2014
Th cells promote CTL survival and memory via acquired pMHC-I and endogenous IL-2 and CD40L signaling and by modulating apoptosis-controlling pathways.
PloS one
Umeshappa CS,Xie Y,Xu S,Nanjundappa RH,Freywald A,Deng Y,Ma H,Xiang J
13-1271 was used in Flow cytometry/Cell sorting to assess the role of CD4 helper T cells with acquired antigen-presenting machineries in determining cytotoxic T lymphocyte fates.
Tue Jan 21 00:00:00 EST 2014
Clonal analysis unveils self-renewing lineage-restricted progenitors generated directly from hematopoietic stem cells.
Cell
Yamamoto R,Morita Y,Ooehara J,Hamanaka S,Onodera M,Rudolph KL,Ema H,Nakauchi H
13-1271 was used in Flow cytometry/Cell sorting to identify multipotent progenitors of myeloid cells and study differentiation pathways giving rise to common myeloid cells.
Thu Aug 29 00:00:00 EDT 2013
Th1/Th17-mediated interstitial pneumonia in chronic colitis mice independent of intestinal microbiota.
Journal of immunology (Baltimore, Md. : 1950)
Nemoto Y,Kanai T,Takahara M,Oshima S,Okamoto R,Tsuchiya K,Matsumoto S,Watanabe M
13-1271 was used in Flow cytometry/Cell sorting to investigate the role of intestinal microbiota in the development of colitis and extraintestinal manifestations in a mouse model.
Sat Jun 15 00:00:00 EDT 2013
Peyer's patch inducer cells play a leading role in the formation of B and T cell zone architecture.
Journal of immunology (Baltimore, Md. : 1950)
Nakagawa R,Togawa A,Nagasawa T,Nishikawa S
13-1271 was used in Immunohistochemistry to analyse evidence that CD3(-)IL-7Rα(+) PP inducer cells are involved in the formation of B and T cell zones in neonatal mice.
Mon Apr 01 00:00:00 EDT 2013
Coactivator-associated arginine methyltransferase 1 regulates fetal hematopoiesis and thymocyte development.
Journal of immunology (Baltimore, Md. : 1950)
Li J,Zhao Z,Carter C,Ehrlich LI,Bedford MT,Richie ER
13-1271 was used in Flow cytometry/Cell sorting to demonstrate how CARM1 is a key epigenetic regulator of hematopoiesis that affects multiple lineages at various stages of differentiation.
Tue Jan 15 00:00:00 EST 2013
IL-2 and IL-7 determine the homeostatic balance between the regulatory and conventional CD4+ T cell compartments during peripheral T cell reconstitution.
Journal of immunology (Baltimore, Md. : 1950)
Le Campion A,Pommier A,Delpoux A,Stouvenel L,Auffray C,Martin B,Lucas B
13-1271 was used in Flow cytometry/Cell sorting to suggest that the proliferation of regulatory CD4(+) T cells in response to lymphopenia appears to be primarily controlled by IL-2.
Mon Oct 01 00:00:00 EDT 2012
Thymocyte responsiveness to endogenous glucocorticoids is required for immunological fitness.
The Journal of clinical investigation
Mittelstadt PR,Monteiro JP,Ashwell JD
13-1271 was used in Flow cytometry/Cell sorting to assess how endogenous glucocorticoids might influence the adaptive immune response.
Sun Jul 01 00:00:00 EDT 2012
The development of colitogenic CD4(+) T cells is regulated by IL-7 in collaboration with NK cell function in a murine model of colitis.
Journal of immunology (Baltimore, Md. : 1950)
Yamaji O,Nagaishi T,Totsuka T,Onizawa M,Suzuki M,Tsuge N,Hasegawa A,Okamoto R,Tsuchiya K,Nakamura T,Arase H,Kanai T,Watanabe M
13-1271 was used in Flow cytometry/Cell sorting to suggest that NK cells suppress colitis severity in T cell-reconstituted RAG(-/-) and IL-7(-/-)RAG(-/-) recipient mice.
Thu Mar 15 00:00:00 EDT 2012
A self-reinforcing regulatory network triggered by limiting IL-7 activates pre-BCR signaling and differentiation.
Nature immunology
Ochiai K,Maienschein-Cline M,Mandal M,Triggs JR,Bertolino E,Sciammas R,Dinner AR,Clark MR,Singh H
Published figure using CD127 monoclonal antibody (Product # 13-1271-82) in Flow Cytometry
Sun Jan 22 00:00:00 EST 2012
Transcription factor RORα is critical for nuocyte development.
Nature immunology
Wong SH,Walker JA,Jolin HE,Drynan LF,Hams E,Camelo A,Barlow JL,Neill DR,Panova V,Koch U,Radtke F,Hardman CS,Hwang YY,Fallon PG,McKenzie AN
13-1271 was used in Flow cytometry/Cell sorting to study how nuocytes arose in the bone marrow and differentiated from common lymphoid progenitors, indicating they are a distinct, previously unknown member of the lymphoid lineage.
Sun Jan 22 00:00:00 EST 2012
The transcriptional coactivator Cbp regulates self-renewal and differentiation in adult hematopoietic stem cells.
Molecular and cellular biology
Chan WI,Hannah RL,Dawson MA,Pridans C,Foster D,Joshi A,Göttgens B,Van Deursen JM,Huntly BJ
13-1271 was used in Flow cytometry/Cell sorting to investigate the role of Cbp in adult HSC maintenance, and the cellular and molecular mechanisms underlying its function.
Thu Dec 01 00:00:00 EST 2011
Chronic exposure to a TLR ligand injures hematopoietic stem cells.
Journal of immunology (Baltimore, Md. : 1950)
Esplin BL,Shimazu T,Welner RS,Garrett KP,Nie L,Zhang Q,Humphrey MB,Yang Q,Borghesi LA,Kincade PW
13-1271 was used in Flow cytometry/Cell sorting to demonstrate that hematopoietic stem cells in chronically LPS-treated animals are functionally compromised.
Sun May 01 00:00:00 EDT 2011
Growth factor independence 1 protects hematopoietic stem cells against apoptosis but also prevents the development of a myeloproliferative-like disease.
Stem cells (Dayton, Ohio)
Khandanpour C,Kosan C,Gaudreau MC,Dührsen U,Hébert J,Zeng H,Möröy T
Tue Feb 01 00:00:00 EST 2011
Interleukin-7-induced Stat-5 acts in synergy with Flt-3 signaling to stimulate expansion of hematopoietic progenitor cells.
The Journal of biological chemistry
Åhsberg J,Tsapogas P,Qian H,Zetterblad J,Zandi S,Månsson R,Jönsson JI,Sigvardsson M
13-1271 was used in Flow cytometry/Cell sorting to suggest that although Il-7 induced Stat-5 phosphorylation, Flt-3L caused activation of the ERK and AKT signaling pathways.
Fri Nov 19 00:00:00 EST 2010
Unexpected role of clathrin adaptor AP-1 in MHC-dependent positive selection of T cells.
Proceedings of the National Academy of Sciences of the United States of America
Alvarez Arias DA,McCarty N,Lu L,Maldonado RA,Shinohara ML,Cantor H
13-1271 was used in Flow cytometry/Cell sorting to study the impact of AP-1 deficiency on thymocyte maturation and interactions with antigen presenting cells.
Tue Feb 09 00:00:00 EST 2010
Langerhans cell (LC) proliferation mediates neonatal development, homeostasis, and inflammation-associated expansion of the epidermal LC network.
The Journal of experimental medicine
Chorro L,Sarde A,Li M,Woollard KJ,Chambon P,Malissen B,Kissenpfennig A,Barbaroux JB,Groves R,Geissmann F
13-1271 was used in Flow cytometry/Cell sorting to investigate the mechanisms involved in Langerhans cell development and homeostasis.
Mon Dec 21 00:00:00 EST 2009
The transcription factors STAT5A/B regulate GM-CSF-mediated granulopoiesis.
Blood
Kimura A,Rieger MA,Simone JM,Chen W,Wickre MC,Zhu BM,Hoppe PS,O'Shea JJ,Schroeder T,Hennighausen L
13-1271 was used in Flow cytometry/Cell sorting to assess the contribution of the downstream mediating transcription factors STAT5A/B to the development of granulocytes.
Thu Nov 19 00:00:00 EST 2009
The survival of memory CD4+ T cells within the gut lamina propria requires OX40 and CD30 signals.
Journal of immunology (Baltimore, Md. : 1950)
Withers DR,Jaensson E,Gaspal F,McConnell FM,Eksteen B,Anderson G,Agace WW,Lane PJL
13-1271 was used in Flow cytometry/Cell sorting to investigate how signals through CD30 and OX40 may be required for the survival of CD4+ T cells within the gut lamina propria.
Thu Oct 15 00:00:00 EDT 2009
Long-lived colitogenic CD4+ memory T cells residing outside the intestine participate in the perpetuation of chronic colitis.
Journal of immunology (Baltimore, Md. : 1950)
Nemoto Y,Kanai T,Kameyama K,Shinohara T,Sakamoto N,Totsuka T,Okamoto R,Tsuchiya K,Nakamura T,Sudo T,Matsumoto S,Watanabe M
13-1271 was used in Flow cytometry/Cell sorting to investigate the latency of colitogenic CD4 T-cells in the remission stage of colitis under germ-free conditions.
Thu Oct 15 00:00:00 EDT 2009
LTbetaR signaling induces cytokine expression and up-regulates lymphangiogenic factors in lymph node anlagen.
Journal of immunology (Baltimore, Md. : 1950)
Vondenhoff MF,Greuter M,Goverse G,Elewaut D,Dewint P,Ware CF,Hoorweg K,Kraal G,Mebius RE
Fri May 01 00:00:00 EDT 2009
The role of thymic stromal lymphopoietin in CD8+ T cell homeostasis.
Journal of immunology (Baltimore, Md. : 1950)
Rochman Y,Leonard WJ
13-1271 was used in Flow cytometry/Cell sorting to elucidate whether the thymic stromal lymphopoietin receptor component, the IL-7R alpha chain, contributes to CD8(+) T cell survival.
Mon Dec 01 00:00:00 EST 2008
IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Journal of immunology (Baltimore, Md. : 1950)
Kikuchi K,Kasai H,Watanabe A,Lai AY,Kondo M
13-1271 was used in Flow cytometry/Cell sorting to investigate when IL-7 stimulation is necessary in hemato-/lymphopoiesis in adult mice, showing that IL-7 specifies B cell fate at the common lymphoid progenitor to pre-proB transition stage by maintaining early B cell factor expression.
Tue Jul 01 00:00:00 EDT 2008
Characterization of developmental pathway of natural killer cells from embryonic stem cells in vitro.
PloS one
Tabatabaei-Zavareh N,Vlasova A,Greenwood CP,Takei F
13-1271 was used in Flow cytometry/Cell sorting to study lymphocyte development from embryonic stem cells and characterise natural killer cell developmental intermediates using a multi-step culture system.
Wed Feb 21 00:00:00 EST 2007
Msx2-interacting nuclear target protein (Mint) deficiency reveals negative regulation of early thymocyte differentiation by Notch/RBP-J signaling.
Proceedings of the National Academy of Sciences of the United States of America
Tsuji M,Shinkura R,Kuroda K,Yabe D,Honjo T
13-1271-82 was used in Flow Cytometry to o investigate where and how Notch regulates early T cell differentiation by using specific up-regulation of Notch signaling by Mint knockout.
Tue Jan 30 00:00:00 EST 2007
The order of expression of transcription factors directs hierarchical specification of hematopoietic lineages.
Genes & development
Iwasaki H,Mizuno S,Arinobu Y,Ozawa H,Mori Y,Shigematsu H,Takatsu K,Tenen DG,Akashi K
13-1271 was used in Flow cytometry/Cell sorting to show that the expression timing of GATA-2 and CCAAT enhancer-binding protein alpha can differentially control their lineage commitment.
Wed Nov 01 00:00:00 EST 2006
Proper levels of c-Myb are discretely defined at distinct steps of hematopoietic cell development.
Blood
Sakamoto H,Dai G,Tsujino K,Hashimoto K,Huang X,Fujimoto T,Mucenski M,Frampton J,Ogawa M
13-1271 was used in Flow cytometry/Cell sorting to investigate the functional role of c-Myb in hematopoietic cell development from endothelial cells, showing that proper levels of c-Myb are discretely defined at distinct steps of development.
Tue Aug 01 00:00:00 EDT 2006
Foxp3-expressing CD103+ regulatory T cells accumulate in dendritic cell aggregates of the colonic mucosa in murine transfer colitis.
The American journal of pathology
Leithäuser F,Meinhardt-Krajina T,Fink K,Wotschke B,Möller P,Reimann J
Thu Jun 01 00:00:00 EDT 2006
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
13-1271 was used in Flow cytometry/Cell sorting to investigate the control of haematopoiesis by peroxide-sensitive regulatory mechanisms.
Wed Mar 01 00:00:00 EST 2006
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
13-1271 was used in Flow cytometry/Cell sorting to investigate the control of haematopoiesis by peroxide-sensitive regulatory mechanisms.
Wed Mar 01 00:00:00 EST 2006
Distinctive and indispensable roles of PU.1 in maintenance of hematopoietic stem cells and their differentiation.
Blood
Iwasaki H,Somoza C,Shigematsu H,Duprez EA,Iwasaki-Arai J,Mizuno S,Arinobu Y,Geary K,Zhang P,Dayaram T,Fenyus ML,Elf S,Chan S,Kastner P,Huettner CS,Murray R,Tenen DG,Akashi K
13-1271 was used in Flow cytometry/Cell sorting to show that constitutive expression of PU.1 by HSCs is necessary for maintenance of the bone marrow HSC pool.
Thu Sep 01 00:00:00 EDT 2005
T/B lineage choice occurs prior to intrathymic Notch signaling.
Blood
Harman BC,Jenkinson WE,Parnell SM,Rossi SW,Jenkinson EJ,Anderson G
13-1271 was used in Flow cytometry/Cell sorting to analyse the T-cell/B-cell lineage potential of precursors that have been recruited to the foetal mouse thymus, showing that lineage choice occurs prior to intrathymic Notch signaling.
Mon Aug 01 00:00:00 EDT 2005
Increased expression of germinal center-associated nuclear protein RNA-primase is associated with lymphomagenesis.
Cancer research
Fujimura S,Xing Y,Takeya M,Yamashita Y,Ohshima K,Kuwahara K,Sakaguchi N
13-1271 was used in Flow cytometry/Cell sorting to study the role of GANP in lymphomagenesis.
Fri Jul 01 00:00:00 EDT 2005
Hmgb3: an HMG-box family member expressed in primitive hematopoietic cells that inhibits myeloid and B-cell differentiation.
Blood
Nemeth MJ,Curtis DJ,Kirby MR,Garrett-Beal LJ,Seidel NE,Cline AP,Bodine DM
13-1271 was used in Flow cytometry/Cell sorting to elucidate the role of Hmgb3 regulation in myeloid and B-cell differentiation.
Fri Aug 15 00:00:00 EDT 2003