Search Thermo Fisher Scientific
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
Mouse anti Human CD173 antibody, clone BRIC231 recognizes human type 2 H blood group antigen, also known as CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes.
CD173 is also known as human Type 2 H blood group antigen. Active H substances are expressed by many cells and tissues, including erythrocytes.
Protein Aliases: H2; Type 2 H blood group antigen