Product References
Polycomb Factor PHF19 Controls Cell Growth and Differentiation Toward Erythroid Pathway in Chronic Myeloid Leukemia Cells.
Frontiers in cell and developmental biology
García-Montolio M,Ballaré C,Blanco E,Gutiérrez A,Aranda S,Gómez A,Kok CH,Yeung DT,Hughes TP,Vizán P,Di Croce L
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Tue May 18 00:00:00 EDT 2021
Diurnal Variation of Plasma Extracellular Vesicle Is Disrupted in People Living with HIV.
Pathogens (Basel, Switzerland)
Bazié WW,Goyer B,Boucher J,Zhang Y,Planas D,Chatterjee D,Routy JP,Alary M,Ancuta P,Gilbert C
11-9987-82 was used in Flow Cytometry to investigate daily variations of large and small EVs, in terms of abundance and microRNA contents in people living with HIV (PLWH) receiving antiretroviral therapy (HIV+ART) and uninfected controls (HIV-).
Sat Apr 24 00:00:00 EDT 2021
Iron control of erythroid microtubule cytoskeleton as a potential target in treatment of iron-restricted anemia.
Nature communications
Goldfarb AN,Freeman KC,Sahu RK,Elagib KE,Holy M,Arneja A,Polanowska-Grabowska R,Gru AA,White Z,Khalil S,Kerins MJ,Ooi A,Leitinger N,Luckey CJ,Delehanty LL
11-9987 was used in Flow cytometry/Cell sorting to identify a cytoskeletal component of erythroid iron restriction and demonstrate potential for its therapeutic targeting in ACDI.
Fri Mar 12 00:00:00 EST 2021
Iron control of erythroid microtubule cytoskeleton as a potential target in treatment of iron-restricted anemia.
Nature communications
Goldfarb AN,Freeman KC,Sahu RK,Elagib KE,Holy M,Arneja A,Polanowska-Grabowska R,Gru AA,White Z,Khalil S,Kerins MJ,Ooi A,Leitinger N,Luckey CJ,Delehanty LL
11-9987 was used in Flow cytometry/Cell sorting to identify a cytoskeletal component of erythroid iron restriction and demonstrate potential for its therapeutic targeting in ACDI.
Fri Mar 12 00:00:00 EST 2021
Heterogeneous disease-propagating stem cells in juvenile myelomonocytic leukemia.
The Journal of experimental medicine
Louka E,Povinelli B,Rodriguez-Meira A,Buck G,Wen WX,Wang G,Sousos N,Ashley N,Hamblin A,Booth CAG,Roy A,Elliott N,Iskander D,de la Fuente J,Fordham N,O'Byrne S,Inglott S,Norfo R,Salio M,Thongjuea S,Rao A,Roberts I,Mead AJ
11-9987-82 was used in Flow Cytometry to pave the way for LSC-directed disease monitoring and therapy in this disease.
Mon Feb 01 00:00:00 EST 2021
A Human Skeletal Muscle Atlas Identifies the Trajectories of Stem and Progenitor Cells across Development and from Human Pluripotent Stem Cells.
Cell stem cell
Xi H,Langerman J,Sabri S,Chien P,Young CS,Younesi S,Hicks M,Gonzalez K,Fujiwara W,Marzi J,Liebscher S,Spencer M,Van Handel B,Evseenko D,Schenke-Layland K,Plath K,Pyle AD
11-9987-82 was used in Flow Cytometry to profile human embryonic, fetal and postnatal skeletal muscle tissues to develop a roadmap of muscle ontogeny.
Thu Jul 02 00:00:00 EDT 2020
Gene-centric functional dissection of human genetic variation uncovers regulators of hematopoiesis.
eLife
Nandakumar SK,McFarland SK,Mateyka LM,Lareau CA,Ulirsch JC,Ludwig LS,Agarwal G,Engreitz JM,Przychodzen B,McConkey M,Cowley GS,Doench JG,Maciejewski JP,Ebert BL,Root DE,Sankaran VG
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Thu May 09 00:00:00 EDT 2019
Genetic programming of macrophages generates an in vitro model for the human erythroid island niche.
Nature communications
Lopez-Yrigoyen M,Yang CT,Fidanza A,Cassetta L,Taylor AH,McCahill A,Sellink E,von Lindern M,van den Akker E,Mountford JC,Pollard JW,Forrester LM
11-9987 was used in Flow cytometry/Cell sorting to develop an in vitro system to model the human erythroblastic island niche.
Wed Feb 20 00:00:00 EST 2019
Mechanisms of establishment and functional significance of DNA demethylation during erythroid differentiation.
Blood advances
Bartholdy B,Lajugie J,Yan Z,Zhang S,Mukhopadhyay R,Greally JM,Suzuki M,Bouhassira EE
11-9987 was used in Flow cytometry/Cell sorting to study the mechanisms of establishment and functional significance of DNA demethylation during erythroid differentiation.
Tue Aug 14 00:00:00 EDT 2018
Acquired Resistance to FGFR Inhibitor in Diffuse-Type Gastric Cancer through an AKT-Independent PKC-Mediated Phosphorylation of GSK3β.
Molecular cancer therapeutics
Lau WM,Teng E,Huang KK,Tan JW,Das K,Zang Z,Chia T,Teh M,Kono K,Yong WP,Shabbir A,Tay A,Phua NS,Tan P,Chan SL,So JBY
Mon Jan 01 00:00:00 EST 2018
Activation of KLF1 Enhances the Differentiation and Maturation of Red Blood Cells from Human Pluripotent Stem Cells.
Stem cells (Dayton, Ohio)
Yang CT,Ma R,Axton RA,Jackson M,Taylor AH,Fidanza A,Marenah L,Frayne J,Mountford JC,Forrester LM
11-9987 was used in Flow cytometry/Cell sorting to investigate the role of KLF1 in maturation of red blood cells.
Sat Apr 01 00:00:00 EDT 2017
Platelets confound the measurement of extracellular miRNA in archived plasma.
Scientific reports
Mitchell AJ,Gray WD,Hayek SS,Ko YA,Thomas S,Rooney K,Awad M,Roback JD,Quyyumi A,Searles CD
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Tue Sep 13 00:00:00 EDT 2016
Genetic Variability Overrides the Impact of Parental Cell Type and Determines iPSC Differentiation Potential.
Stem cell reports
Kyttälä A,Moraghebi R,Valensisi C,Kettunen J,Andrus C,Pasumarthy KK,Nakanishi M,Nishimura K,Ohtaka M,Weltner J,Van Handel B,Parkkonen O,Sinisalo J,Jalanko A,Hawkins RD,Woods NB,Otonkoski T,Trokovic R
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Tue Feb 09 00:00:00 EST 2016
PPAR-α and glucocorticoid receptor synergize to promote erythroid progenitor self-renewal.
Nature
Lee HY,Gao X,Barrasa MI,Li H,Elmes RR,Peters LL,Lodish HF
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Thu Jun 25 00:00:00 EDT 2015
Allele-specific analysis of DNA replication origins in mammalian cells.
Nature communications
Bartholdy B,Mukhopadhyay R,Lajugie J,Aladjem MI,Bouhassira EE
11-9987 was used in Flow cytometry/Cell sorting to delineate the mechanisms that control the location and timing of firing of replication origins.
Tue May 19 00:00:00 EDT 2015
EDAG positively regulates erythroid differentiation and modifies GATA1 acetylation through recruiting p300.
Stem cells (Dayton, Ohio)
Zheng WW,Dong XM,Yin RH,Xu FF,Ning HM,Zhang MJ,Xu CW,Yang Y,Ding YL,Wang ZD,Zhao WB,Tang LJ,Chen H,Wang XH,Zhan YQ,Yu M,Ge CH,Li CY,Yang XM
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Altered translation of GATA1 in Diamond-Blackfan anemia.
Nature medicine
Ludwig LS,Gazda HT,Eng JC,Eichhorn SW,Thiru P,Ghazvinian R,George TI,Gotlib JR,Beggs AH,Sieff CA,Lodish HF,Lander ES,Sankaran VG
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Tue Jul 01 00:00:00 EDT 2014
Cutting-edge analysis of extracellular microparticles using ImageStream(X) imaging flow cytometry.
Scientific reports
Headland SE,Jones HR,D'Sa AS,Perretti M,Norling LV
11-9987 was used in Immunocytochemistry to investigate methods to quantify microparticle levels.
Tue Jun 10 00:00:00 EDT 2014
A feedback loop consisting of microRNA 23a/27a and the β-like globin suppressors KLF3 and SP1 regulates globin gene expression.
Molecular and cellular biology
Ma Y,Wang B,Jiang F,Wang D,Liu H,Yan Y,Dong H,Wang F,Gong B,Zhu Y,Dong L,Yin H,Zhang Z,Zhao H,Wu Z,Zhang J,Zhou J,Yu J
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
α-1,6-Fucosyltransferase (FUT8) inhibits hemoglobin production during differentiation of murine and K562 human erythroleukemia cells.
The Journal of biological chemistry
Sasaki H,Toda T,Furukawa T,Mawatari Y,Takaesu R,Shimizu M,Wada R,Kato D,Utsugi T,Ohtsu M,Murakami Y
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Fri Jun 07 00:00:00 EDT 2013
Terminal differentiation and loss of tumorigenicity of human cancers via pluripotency-based reprogramming.
Oncogene
Zhang X,Cruz FD,Terry M,Remotti F,Matushansky I
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Immunofluorescence
Thu May 02 00:00:00 EDT 2013
Erythroid-specific expression of β-globin from Sleeping Beauty-transduced human hematopoietic progenitor cells.
PloS one
Sjeklocha LM,Park CW,Wong PY,Roney MJ,Belcher JD,Kaufman DS,Vercellotti GM,Hebbel RP,Steer CJ
11-9987 was used in Flow cytometry/Cell sorting to investigate methods for efficient delivery of tightly regulated and stably expressed gene products, showing erythroid-specific expression of β-globin using the Sleeping Beauty transposon system.
Mon May 21 00:00:00 EDT 2012
Role of ZBP-89 in human globin gene regulation and erythroid differentiation.
Blood
Woo AJ,Kim J,Xu J,Huang H,Cantor AB
Published figure using CD235a (Glycophorin A) monoclonal antibody (Product # 11-9987-82) in Flow Cytometry
Thu Sep 29 00:00:00 EDT 2011
CpG and non-CpG oligodeoxynucleotides directly costimulate mouse and human CD4+ T cells through a TLR9- and MyD88-independent mechanism.
Journal of immunology (Baltimore, Md. : 1950)
Landrigan A,Wong MT,Utz PJ
11-9987 was used in Flow cytometry/Cell sorting to study the ability of oligodeoxynucleotides to activate and polarise T cells.
Thu Sep 15 00:00:00 EDT 2011
Binding and uptake of H-ferritin are mediated by human transferrin receptor-1.
Proceedings of the National Academy of Sciences of the United States of America
Li L,Fang CJ,Ryan JC,Niemi EC,Lebrón JA,Björkman PJ,Arase H,Torti FM,Torti SV,Nakamura MC,Seaman WE
11-9987 was used in Flow cytometry/Cell sorting to demonstrate that TfR1 can bind HFt as well as Tf.
Tue Feb 23 00:00:00 EST 2010