Search Thermo Fisher Scientific
In direct ELISAs, no cross-reactivity with recombinant mouse (rm) LMIR4, rmCD300b/LMIR5, recombinant human (rh) CD300a/LMIR1, rhCD300c/LMIR2, rhCD300f/LMIR3, or rhCD300e/LMIR6 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.