Product References
A combination therapy of bortezomib, CXCR4 inhibitor, and checkpoint inhibitor is effective in cholangiocarcinoma in vivo.
iScience
Li L,Zhou Y,Zhang Y,Hu H,Mao HQ,Selaru FM
14-9326-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to find that the inhibition of CXCR4 allowed bortezomib to sensitize CCA to anti-PD1 treatment, with a significant reduction of tumor burden and long-term overall survival.
Fri Mar 17 00:00:00 EDT 2023
Dynamics of CXCR4 positive circulating tumor cells in prostate cancer patients during radiotherapy.
International journal of cancer
Klusa D,Lohaus F,Franken A,Baumbach M,Cojoc M,Dowling P,Linge A,Offermann A,Löck S,Hušman D,Rivandi M,Polzer B,Freytag V,Lange T,Neubauer H,Kücken M,Perner S,Hölscher T,Dubrovska A,Krause M,Kurth I,Baumann M,Peitzsch C
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Thu Jun 15 00:00:00 EDT 2023
Improved platform for breast cancer circulating tumor cell enrichment and characterization with next-generation sequencing technology.
American journal of cancer research
Mihalcioiu C,Li J,Badescu D,Camirand A,Kremer N,Bertos N,Omeroglu A,Sebag M,Di Battista J,Park M,Ragoussis J,Kremer R
14-9326-82 was used in Immunocytochemistry-immunofluorescence to develop a platform based on the large-scale isolation of CTCs by apheresis, allowing us to collect CTCs in large numbers, which were preserved live in liquid nitrogen for further characterization.
Tue Feb 14 00:00:00 EST 2023
The sodium/glucose cotransporters as potential therapeutic targets for CF lung diseases revealed by human lung organoid swelling assay.
Molecular therapy. Methods & clinical development
Hirai H,Liang X,Sun Y,Zhang Y,Zhang J,Chen YE,Mou H,Zhao Y,Xu J
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Thu Mar 10 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
Tumor suppressor DEAR1 regulates mammary epithelial cell fate and predicts early onset and metastasis in triple negative breast cancer.
Scientific reports
Le UQ,Chen N,Balasenthil S,Lurie E,Yang F,Liu S,Rubin L,Solis Soto LM,Raso MG,Batra H,Sahin AA,Wistuba II,Killary AM
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Mon Nov 14 00:00:00 EST 2022
The Effect of Infant Gastric Digestion on Human Maternal Milk Cells.
Molecular nutrition & food research
Doerfler R,Melamed JR,Whitehead KA
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Sat Oct 01 00:00:00 EDT 2022
Parathyroid Hormone-Related Protein Inhibition Blocks Triple-Negative Breast Cancer Expansion in Bone Through Epithelial to Mesenchymal Transition Reversal.
JBMR plus
Li J,Camirand A,Zakikhani M,Sellin K,Guo Y,Luan X,Mihalcioiu C,Kremer R
14-9326-82 was used in Immunocytochemistry-immunoflourescence to conclude that PTHrP is a targetable EMT molecular driver and suggest that its pharmacological blockade can provide a potential therapeutic approach against established TNBC-derived skeletal lesions.
Wed Jun 01 00:00:00 EDT 2022
Selection of Cancer Stem Cell-Targeting Agents Using Bacteriophage Display.
Methods in molecular biology (Clifton, N.J.)
Prater AR,Deutscher SL
14-9326-82 was used in Flow cytometry/Cell sorting to discuss in detail how to isolate a CSC like population of cells from an aggressive cancer cell line, perform in vivo and in vitro phage display selections against the CSCs, and then characterize the resulting phage/peptides for further use as a diagnostic and therapeutic tool.
Wed Mar 30 00:00:00 EDT 2022
GSDMB is increased in IBD and regulates epithelial restitution/repair independent of pyroptosis.
Cell
Rana N,Privitera G,Kondolf HC,Bulek K,Lechuga S,De Salvo C,Corridoni D,Antanaviciute A,Maywald RL,Hurtado AM,Zhao J,Huang EH,Li X,Chan ER,Simmons A,Bamias G,Abbott DW,Heaney JD,Ivanov AI,Pizarro TT
14-9326-82 was used in Immunohistochemistry to reveal lack of inherent GSDMB-dependent pyroptosis in activated epithelial cells and organoids but instead point to increased proliferation and migration during in vitro wound closure, which arrests in GSDMB-deficient cells that display hyper-adhesiveness and enhanced formation of vinculin-based focal adhesions dependent on PDGF-A-mediated FAK phosphorylation.
Thu Jan 20 00:00:00 EST 2022
Human Nasal and Lung Tissues Infected Ex Vivo with SARS-CoV-2 Provide Insights into Differential Tissue-Specific and Virus-Specific Innate Immune Responses in the Upper and Lower Respiratory Tract.
Journal of virology
Alfi O,Yakirevitch A,Wald O,Wandel O,Izhar U,Oiknine-Djian E,Nevo Y,Elgavish S,Dagan E,Madgar O,Feinmesser G,Pikarsky E,Bronstein M,Vorontsov O,Jonas W,Ives J,Walter J,Zakay-Rones Z,Oberbaum M,Panet A,Wolf DG
14-9326-82 was used in Immunohistochemistry to investigate viral susceptibility and early patterns of local mucosal innate immune response in the authentic milieu of the human respiratory tract. SARS-CoV-2 productively infected the nasal turbinate tissues, predominantly targeting respiratory epithelial cells, with a rapid increase in tissue-associated viral subgenomic mRNA and secretion of infectious viral progeny.
Thu Jun 24 00:00:00 EDT 2021
EMP3 negatively modulates breast cancer cell DNA replication, DNA damage repair, and stem-like properties.
Cell death & disease
Zhou K,Sun Y,Dong D,Zhao C,Wang W
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Sun Sep 12 00:00:00 EDT 2021
Helicobacter pylori promote inflammation and host defense through the cagA-dependent activation of mTORC1.
Journal of cellular physiology
Feng GJ,Chen Y,Li K
14-9326-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to support H. pylori promote gastritis and inhibit bacterial colonization through the cagA-dependent activation of mTORC1.
Tue Dec 01 00:00:00 EST 2020
Helicobacter pylori promote inflammation and host defense through the cagA-dependent activation of mTORC1.
Journal of cellular physiology
Feng GJ,Chen Y,Li K
14-9326-82 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to support H. pylori promote gastritis and inhibit bacterial colonization through the cagA-dependent activation of mTORC1.
Tue Dec 01 00:00:00 EST 2020
FZD5 contributes to TNBC proliferation, DNA damage repair and stemness.
Cell death & disease
Sun Y,Wang Z,Na L,Dong D,Wang W,Zhao C
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Sat Dec 12 00:00:00 EST 2020
Chemotherapeutic Stress Influences Epithelial-Mesenchymal Transition and Stemness in Cancer Stem Cells of Triple-Negative Breast Cancer.
International journal of molecular sciences
Li X,Strietz J,Bleilevens A,Stickeler E,Maurer J
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Profiling human breast epithelial cells using single cell RNA sequencing identifies cell diversity.
Nature communications
Nguyen QH,Pervolarakis N,Blake K,Ma D,Davis RT,James N,Phung AT,Willey E,Kumar R,Jabart E,Driver I,Rock J,Goga A,Khan SA,Lawson DA,Werb Z,Kessenbrock K
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Wed May 23 00:00:00 EDT 2018
TGF-β1 secreted by Tregs in lymph nodes promotes breast cancer malignancy via up-regulation of IL-17RB.
EMBO molecular medicine
Huang SC,Wei PC,Hwang-Verslues WW,Kuo WH,Jeng YM,Hu CM,Shew JY,Huang CS,Chang KJ,Lee EY,Lee WH
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
A genome editing approach to study cancer stem cells in human tumors.
EMBO molecular medicine
Cortina C,Turon G,Stork D,Hernando-Momblona X,Sevillano M,Aguilera M,Tosi S,Merlos-Suárez A,Stephan-Otto Attolini C,Sancho E,Batlle E
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Sat Jul 01 00:00:00 EDT 2017
Evidence of In Vitro Preservation of Human Nephrogenesis at the Single-Cell Level.
Stem cell reports
Pode-Shakked N,Gershon R,Tam G,Omer D,Gnatek Y,Kanter I,Oriel S,Katz G,Harari-Steinberg O,Kalisky T,Dekel B
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2017
Evidence of In Vitro Preservation of Human Nephrogenesis at the Single-Cell Level.
Stem cell reports
Pode-Shakked N,Gershon R,Tam G,Omer D,Gnatek Y,Kanter I,Oriel S,Katz G,Harari-Steinberg O,Kalisky T,Dekel B
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2017
Triple-negative breast cancer targeting and killing by EpCAM-directed, plasmonically active nanodrug systems.
NPJ precision oncology
Jenkins SV,Nima ZA,Vang KB,Kannarpady G,Nedosekin DA,Zharov VP,Griffin RJ,Biris AS,Dings RPM
14-9326 was used in Flow cytometry/Cell sorting to engineer a nanodrug delivery system consisting of silver-coated golf nanorods targeting epithelial cell adhesion/activating molecule and loaded with doxorubicin.
Thu Oct 01 00:00:00 EDT 2020
Stage-specific embryonic antigen-3 (SSEA-3) and β3GalT5 are cancer specific and significant markers for breast cancer stem cells.
Proceedings of the National Academy of Sciences of the United States of America
Cheung SK,Chuang PK,Huang HW,Hwang-Verslues WW,Cho CH,Yang WB,Shen CN,Hsiao M,Hsu TL,Chang CF,Wong CH
14932682 was used in flow cytometry to find that breast cancer stem cells isolated with CD44(+)CD24(-/lo)SSEA-3(+) or ESA(hi)PROCR(hi)SSEA-3(+) markers have higher tumorigenicity than those with conventional markers
Tue Jan 26 00:00:00 EST 2016
Clostridium perfringens enterotoxin C-terminal domain labeled to fluorescent dyes for in vivo visualization of micrometastatic chemotherapy-resistant ovarian cancer.
International journal of cancer
Cocco E,Shapiro EM,Gasparrini S,Lopez S,Schwab CL,Bellone S,Bortolomai I,Sumi NJ,Bonazzoli E,Nicoletti R,Deng Y,Saltzman WM,Zeiss CJ,Centritto F,Black JD,Silasi DA,Ratner E,Azodi M,Rutherford TJ,Schwartz PE,Pecorelli S,Santin AD
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Immunofluorescence
Tue Dec 01 00:00:00 EST 2015
The isolation and characterization of CTC subsets related to breast cancer dormancy.
Scientific reports
Vishnoi M,Peddibhotla S,Yin W,T Scamardo A,George GC,Hong DS,Marchetti D
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Thu Dec 03 00:00:00 EST 2015
A lung-on-a-chip array with an integrated bio-inspired respiration mechanism.
Lab on a chip
Stucki AO,Stucki JD,Hall SR,Felder M,Mermoud Y,Schmid RA,Geiser T,Guenat OT
14-9326 was used in Flow cytometry/Cell sorting to report a novel lung-on-a-chip that mimics the pulmonary parenchymal environment, including the alveolar barrier and the 3D cyclic strain induced by breathing movements.
Sat Mar 07 00:00:00 EST 2015
NDY1/KDM2B functions as a master regulator of polycomb complexes and controls self-renewal of breast cancer stem cells.
Cancer research
Kottakis F,Foltopoulou P,Sanidas I,Keller P,Wronski A,Dake BT,Ezell SA,Shen Z,Naber SP,Hinds PW,McNiel E,Kuperwasser C,Tsichlis PN
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Tue Jul 15 00:00:00 EDT 2014
GATA3 suppresses metastasis and modulates the tumour microenvironment by regulating microRNA-29b expression.
Nature cell biology
Chou J,Lin JH,Brenot A,Kim JW,Provot S,Werb Z
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013
Estrogen and progesterone together expand murine endometrial epithelial progenitor cells.
Stem cells (Dayton, Ohio)
Janzen DM,Cheng D,Schafenacker AM,Paik DY,Goldstein AS,Witte ON,Jaroszewicz A,Pellegrini M,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Immunohistochemistry
Mon Apr 01 00:00:00 EDT 2013
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Immunofluorescence
Thu Nov 01 00:00:00 EDT 2012
Stem-like epithelial cells are concentrated in the distal end of the fallopian tube: a site for injury and serous cancer initiation.
Stem cells (Dayton, Ohio)
Paik DY,Janzen DM,Schafenacker AM,Velasco VS,Shung MS,Cheng D,Huang J,Witte ON,Memarzadeh S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Immunofluorescence
Thu Nov 01 00:00:00 EDT 2012
Slow-cycling therapy-resistant cancer cells.
Stem cells and development
Moore N,Houghton J,Lyle S
Published figure using CD326 (EpCAM) monoclonal antibody (Product # 14-9326-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012