Product References
Endovascular mechanical thrombectomy in a child with COVID-19: Clot analysis reveals a novel pathway in the neuroinflammatory cascade resulting in large-vessel occlusion.
Interventional neuroradiology : journal of peritherapeutic neuroradiology, surgical procedures and related neurosciences
Ravindra VM,Denorme F,D Alexander M,A Campbell R,Grandhi R
MA5-11642 was used in Flow Cytometry, Immunohistochemistry (Paraffin) to emphasize the importance of recognizing SARS-CoV-2 and the propensity for thrombosis in a delayed fashion, which can lead to severe stroke in young people.
Sun Oct 01 00:00:00 EDT 2023
Clot composition characterization using diffuse reflectance spectroscopy in acute ischemic stroke.
Biomedical optics express
Skyrman S,Burström G,Aspegren O,Babic D,Lucassen G,Edström E,Arnberg F,Ohlsson M,Mueller M,Elmi-Terander A,Andersson T
MA5-11642 was used in Immunocytochemistry-immunoflourescence to show that, applied in an intravascular device, DRS could potentially provide intraprocedural information on clot composition that could increase endovascular thrombectomy efficiency.
Wed Jun 01 00:00:00 EDT 2022
Platelet-Neutrophil Association in NETs-Rich Areas in the Retrieved AIS Patient Thrombi.
International journal of molecular sciences
Pir GJ,Parray A,Ayadathil R,Pananchikkal SV,Mir FA,Muhammad I,Abubakar A,Amir N,Hussain S,Haroon KH,Muhammad A,Imam Y,Patro SN,Akhtar N,Zakaria A,Kamran S
MA5-11642 was used in Flow Cytometry, Immunohistochemistry (Paraffin) to show that NETs-containing areas showed strong reactivity to VWF, platelets, and high-mobility group box 1 (HMGB1), signifying a close interplay between these components.
Mon Nov 21 00:00:00 EST 2022
High Mobility Group Box 1 Protein in Cerebral Thromboemboli.
International journal of molecular sciences
Essig F,Babilon L,Vollmuth C,Kollikowski AM,Pham M,Solymosi L,Haeusler KG,Kraft P,Stoll G,Schuhmann MK
MA5-11642 was used in Immunohistochemistry to demonstrate that HMGB1 is a substantial constituent of thromboemboli causing large vessel occlusion stroke.
Tue Oct 19 00:00:00 EDT 2021
High Mobility Group Box 1 Protein in Cerebral Thromboemboli.
International journal of molecular sciences
Essig F,Babilon L,Vollmuth C,Kollikowski AM,Pham M,Solymosi L,Haeusler KG,Kraft P,Stoll G,Schuhmann MK
MA5-11642 was used in Immunohistochemistry to demonstrate that HMGB1 is a substantial constituent of thromboemboli causing large vessel occlusion stroke.
Tue Oct 19 00:00:00 EDT 2021
Structural analysis of ischemic stroke thrombi: histological indications for therapy resistance.
Haematologica
Staessens S,Denorme F,Francois O,Desender L,Dewaele T,Vanacker P,Deckmyn H,Vanhoorelbeke K,Andersson T,De Meyer SF
MA5-11642 was used in Immunohistochemistry-immunofluorescence to study the internal organisation of ischemic stroke thrombi on a cellular and molecular level.
Tue Apr 27 00:00:00 EDT 2021
Structural analysis of ischemic stroke thrombi: histological indications for therapy resistance.
Haematologica
Staessens S,Denorme F,Francois O,Desender L,Dewaele T,Vanacker P,Deckmyn H,Vanhoorelbeke K,Andersson T,De Meyer SF
MA5-11642 was used in Immunohistochemistry-immunofluorescence to study the internal organisation of ischemic stroke thrombi on a cellular and molecular level.
Tue Apr 27 00:00:00 EDT 2021
Redox control of β2-glycoprotein I-von Willebrand factor interaction by thioredoxin-1.
Journal of thrombosis and haemostasis : JTH
Passam FH,Rahgozar S,Qi M,Raftery MJ,Wong JW,Tanaka K,Ioannou Y,Zhang JY,Gemmell R,Qi JC,Giannakopoulos B,Hughes WE,Hogg PJ,Krilis SA
MA5-11642 was used in immunocytochemistry to investigate the interaction between beta2-GPI and von Willebrand factor and the effect of thioredoxin-1 on this interaction
Sun Aug 01 00:00:00 EDT 2010
Crystal structure of a platelet-agglutinating factor isolated from the venom of Taiwan habu (Trimeresurus mucrosquamatus).
The Biochemical journal
Huang KF,Ko TP,Hung CC,Chu J,Wang AH,Chiou SH
MA5-11642 was used in blocking or activating experiment to determine the crystal structure of the tetrameric platelet aggregating factor mucrocetin purified from snake venom
Mon Mar 01 00:00:00 EST 2004