Product References
Activation of Human Vδ2+ γδ T Cells by Staphylococcus aureus Promotes Enhanced Anti-Staphylococcal Adaptive Immunity.
Journal of immunology (Baltimore, Md. : 1950)
Cooper AJR,Lalor SJ,McLoughlin RM
MA1-33243 was used in Flow Cytometry to study the capacity of human Vδ2+ γδ T cells for rapid activation in response to S. aureus.
Sat Aug 15 00:00:00 EDT 2020
Phase I Study of Ficlatuzumab and Cetuximab in Cetuximab-Resistant, Recurrent/Metastatic Head and Neck Cancer.
Cancers
Bauman JE,Ohr J,Gooding WE,Ferris RL,Duvvuri U,Kim S,Johnson JT,Soloff AC,Wallweber G,Winslow J,Gaither-Davis A,Grandis JR,Stabile LP
MA1-33243 was used in Flow Cytometry to establish the recommended Phase II dose (RP2D). Secondary objectives included overall response rate (ORR), progression-free survival (PFS), and overall survival (OS).
Thu Jun 11 00:00:00 EDT 2020
Allergen-specific immunotherapy modulates the balance of circulating Tfh and Tfr cells.
The Journal of allergy and clinical immunology
Schulten V,Tripple V,Seumois G,Qian Y,Scheuermann RH,Fu Z,Locci M,Rosales S,Vijayanand P,Sette A,Alam R,Crotty S,Peters B
MA1-33243 was used in Flow cytometry/Cell sorting to show balancing Tfh and Tfr cells in circulation can be mediated by allergen-specific immunotherapy.
Thu Feb 01 00:00:00 EST 2018
The absence of HLA class I expression in non-small cell lung cancer correlates with the tumor tissue structure and the pattern of T cell infiltration.
International journal of cancer
Perea F,Bernal M,Sánchez-Palencia A,Carretero J,Torres C,Bayarri C,Gómez-Morales M,Garrido F,Ruiz-Cabello F
MA133243 was used in immunohistochemistry - frozen section to characterize HLA class I-positive and negative tumors
Wed Feb 15 00:00:00 EST 2017
CD103+ intraepithelial T cells in high-grade serous ovarian cancer are phenotypically diverse TCRαβ+ CD8αβ+ T cells that can be targeted for cancer immunotherapy.
Oncotarget
Komdeur FL,Wouters MC,Workel HH,Tijans AM,Terwindt AL,Brunekreeft KL,Plat A,Klip HG,Eggink FA,Leffers N,Helfrich W,Samplonius DF,Bremer E,Wisman GB,Daemen T,Duiker EW,Hollema H,Nijman HW,de Bruyn M
MA133243 was used in flow cytometry to define the contribution of CD103+ tumor-infiltrating lymphocytes to high-grade serous epithelial ovarian cancer
Tue Nov 15 00:00:00 EST 2016
CD30+ extranodal natural killer/T-cell lymphoma mimicking phlegmonous myositis: A case report.
Oncology letters
Yang YJ,Li YX,Liu YB,Yang M,Liu K
MA1-33243 was used in immunohistochemistry (paraffin) to report on a case of CD30-positive extranodal natural killer/T-cell lymphoma.
Thu May 01 00:00:00 EDT 2014
A giant primary sclerosing lipogranuloma of the scrotum.
Turk patoloji dergisi
Mungan S,Karagüzel E,Turan C,Reıs A
MA1-33243 was used in immunohistochemistry to report on a case of giant primary sclerosing lipogranuloma of the scrotum
Tue Jul 08 00:00:00 EDT 2014
Corelation of lymphocytic infiltrates with the prognosis of recurrent colo-rectal cancer.
Chirurgia (Bucharest, Romania : 1990)
Makkai-Popa ST,Luncă S,Dimofte G,Vrânceanu A,Franciug D,Ivanov I,Zugun F,Târcoveanu E,Carasevici E
MA1-33243 was used in immunohistochemistry to study the prognostic value of cytotoxic T-cell infiltrates in patients with recurrent colorectal cancer
Fri Mar 21 00:00:00 EDT 2014
IL-21 can supplement suboptimal Lck-independent MAPK activation in a STAT-3-dependent manner in human CD8(+) T cells.
Journal of immunology (Baltimore, Md. : 1950)
Imataki O,Ansén S,Tanaka M,Butler MO,Berezovskaya A,Milstein MI,Kuzushima K,Nadler LM,Hirano N
MA1-33243 was used in flow cytometry to investigate the role of CD8 and antigen in proliferation of CTLs.
Wed Feb 15 00:00:00 EST 2012
Long-term effects of alemtuzumab on regulatory and memory T-cell subsets in kidney transplantation.
Transplantation
Macedo C,Walters JT,Orkis EA,Isse K,Elinoff BD,Fedorek SP,McMichael JM,Chalasani G,Randhawa P,Demetris AJ,Zeevi A,Tan H,Shapiro R,Landsittel D,Lakkis FG,Metes D
MA1-33243 was used in immunohistochemistry to study the effects on effector and memory T-cell subsets of prolonged treatment with alemtuzumab in kidney transplant patients
Fri Apr 27 00:00:00 EDT 2012
The significance of multiplex PCR/heteroduplex analysis-based TCR-γ gene rearrangement combined with laser-capture microdissection in the diagnosis of early mycosis fungoides.
Journal of cutaneous pathology
Yang H,Xu C,Tang Y,Wan C,Liu W,Wang L
MA1-33243 was used in immunohistochemistry (paraffin) to study the diagnostic value for detecting early mycosis fungoides of a combination of multiplex PCR analyis of TCRgamma gene rearrangements and laser-capture microdissection.
Thu Mar 01 00:00:00 EST 2012
Serous effusion cytology of extranodal natural killer/T-cell lymphoma.
Cytopathology : official journal of the British Society for Clinical Cytology
Su XY,Huang J,Jiang Y,Tang Y,Li GD,Liu WP
MA1-33243 was used in flow cytometry to characterize extranodal natural killer/T-cell lymphoma, nasal type using patient samples.
Sun Apr 01 00:00:00 EDT 2012
Effector Memory T cells Are Associated With Atherosclerosis in Humans and Animal Models.
Journal of the American Heart Association
Ammirati E,Cianflone D,Vecchio V,Banfi M,Vermi AC,De Metrio M,Grigore L,Pellegatta F,Pirillo A,Garlaschelli K,Manfredi AA,Catapano AL,Maseri A,Palini AG,Norata GD
MA1-33243 was used in flow cytometry to study the relationship between circulating CD4+ T-cell subsets and atherosclerosis
Wed Feb 01 00:00:00 EST 2012
Axonal projections of mechanoreceptive dorsal root ganglion neurons depend on Ret.
Development (Cambridge, England)
Honma Y,Kawano M,Kohsaka S,Ogawa M
MA1-33243 was used in immunohistochemistry to study the role of Ret in the neurturin-mediated formation of precise axonal projections in the central nervous system
Thu Jul 01 00:00:00 EDT 2010
A patient with clinicopathologic features of small plaque parapsoriasis presenting later with plaque-stage mycosis fungoides: report of a case and comparative retrospective study of 27 cases of "nonprogressive" small plaque parapsoriasis.
Journal of the American Academy of Dermatology
Belousova IE,Vanecek T,Samtsov AV,Michal M,Kazakov DV
MA1-33243 was used in immunohistochemistry to report on a case of small plaque parapsoriasis with plaque-stage mycosis fungoides
Mon Sep 01 00:00:00 EDT 2008
Hyperplasia of hair follicles and other adnexal structures in cutaneous lymphoproliferative disorders: a study of 53 cases, including so-called pseudolymphomatous folliculitis and overt lymphomas.
The American journal of surgical pathology
Kazakov DV,Belousova IE,Kacerovska D,Sima R,Vanecek T,Vazmitel M,Pizinger K,Michal M
MA1-33243 was used in immunohistochemistry to study 53 cases of cutaneous lymphoproliferative disorders
Wed Oct 01 00:00:00 EDT 2008
Natural regulatory T cells: number and function are normal in the majority of patients with lupus nephritis.
Clinical and experimental immunology
Yates J,Whittington A,Mitchell P,Lechler RI,Lightstone L,Lombardi G
MA1-33243 was used in flow cytometry to examine the frequency, phenotype, and function of T regulatory cells in patients with systemic lupus erythematosus and inactive disease
Tue Jul 01 00:00:00 EDT 2008
Up-regulation of expression of the ubiquitin carboxyl-terminal hydrolase L1 gene in human airway epithelium of cigarette smokers.
Cancer research
Carolan BJ,Heguy A,Harvey BG,Leopold PL,Ferris B,Crystal RG
MA1-33243 was used in immunocytochemistry to investigate the expression of carboxyl-terminal hydrolyase L1 gene in human airway epithelium of cigarette smokers
Wed Nov 15 00:00:00 EST 2006
Monocyte-derived human macrophages mediate anergy in allogeneic T cells and induce regulatory T cells.
Journal of immunology (Baltimore, Md. : 1950)
Hoves S,Krause SW,Schütz C,Halbritter D,Schölmerich J,Herfarth H,Fleck M
MA1-33243 was used in flow cytometry to assess the T cell-stimulatory capacity of mature dendritic cells and macrophages using a model of allogeneic MLR
Tue Aug 15 00:00:00 EDT 2006
Primary cutaneous histiocyte and neutrophil-rich CD30+ and CD56+ anaplastic large-cell lymphoma with prominent angioinvasion and nerve involvement in the forehead and scalp of an immunocompetent woman.
Journal of cutaneous pathology
Boudova L,Kazakov DV,Jindra P,Sima R,Vanecek T,Kuntscher V,Vera V,Bouda J,Michal M
MA1-33243 was used in immunohistochemistry to report on a case of utaneous lymphoma of the head co-expressing CD56 and CD30
Tue Aug 01 00:00:00 EDT 2006
Cutaneous lymphoid hyperplasia and other lymphoid infiltrates of the breast nipple: a retrospective clinicopathologic study of fifty-six patients.
The American Journal of dermatopathology
Boudova L,Kazakov DV,Sima R,Vanecek T,Torlakovic E,Lamovec J,Kutzner H,Szepe P,Plank L,Bouda J,Hes O,Mukensnabl P,Michal M
MA1-33243 was used in immunohistochemistry to study the clinicopathological spectrum of lymphoproliferations and other lymphoid infiltration of the breast nipple
Sat Oct 01 00:00:00 EDT 2005
T-cell-rich B-cell lymphoma: a clinicopathologic study of 21 cases and comparison with 43 cases of diffuse large B-cell lymphoma.
Leukemia research
Aki H,Tuzuner N,Ongoren S,Baslar Z,Soysal T,Ferhanoglu B,Sahinler I,Aydin Y,Ulku B,Aktuglu G
MA1-33243 was used in immunohistochemistry to compare the clinocopathology of T-cell rich B-cell lymphoma and diffuse large B-cell lymphoma
Mon Mar 01 00:00:00 EST 2004
Chemokine receptor CCR4 on CD4+ T cells in juvenile rheumatoid arthritis synovial fluid defines a subset of cells with increased IL-4:IFN-gamma mRNA ratios.
Journal of immunology (Baltimore, Md. : 1950)
Thompson SD,Luyrink LK,Graham TB,Tsoras M,Ryan M,Passo MH,Glass DN
MA1-33243 was used in flow cytometry to elucidate the mechanisms that promote recruitment and survival of T cells within the pediatric inflamed joint.
Fri Jun 01 00:00:00 EDT 2001
Quantification of CD4, CCR5, and CXCR4 levels on lymphocyte subsets, dendritic cells, and differentially conditioned monocyte-derived macrophages.
Proceedings of the National Academy of Sciences of the United States of America
Lee B,Sharron M,Montaner LJ,Weissman D,Doms RW
MA1-33243 was used in flow cytometry to determine the number of CD4, CCR5, and CXCR4 antibody-binding sites on various T cells and macrophages.
Tue Apr 27 00:00:00 EDT 1999
CK beta-11/macrophage inflammatory protein-3 beta/EBI1-ligand chemokine is an efficacious chemoattractant for T and B cells.
Journal of immunology (Baltimore, Md. : 1950)
Kim CH,Pelus LM,White JR,Applebaum E,Johanson K,Broxmeyer HE
MA1-33243 was used in flow cytometry to determine the functional properties of CK beta-11/MIP-3 beta/ELC.
Sun Mar 01 00:00:00 EST 1998
Functional subsets of human helper-inducer cells defined by a new monoclonal antibody, UCHL1.
Immunology
Smith SH,Brown MH,Rowe D,Callard RE,Beverley PC
MA1-33243 was used in flow cytometry, immunohistochemistry - frozen section, and immunoprecipitation to characterize the monoclonal antibody UCHL1.
Thu May 01 00:00:00 EDT 1986