Search Thermo Fisher Scientific
This antibody is a monoclonal mixture of FITC-conjugated CD5 (clone M28623, isotype IgG2a) and PE-conjugated CD19 (clone 1G9, isotype IgG1).
Dual color combination consisting of FITC conjugated and R-PE conjugated monoclonal antibodies mixed in optimal ratio.