Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PFRLSSSLQG SAVTLKTSKM ALSPSATAKR INRTSLSSGA TVRSFSINSR LASSLGNLNA ATDDPENKKT SSSSKAGFTA SPFTNLLNGS SQPTTRNYPE
The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CDC10 (cell division cycle 10, S. cerevisiae, homolog); CDC14 cell division cycle 14 homolog A; Dual specificity protein phosphatase CDC14A
Gene Aliases: cdc14; CDC14A; DFNB105; hCDC14
UniProt ID: (Human) Q9UNH5
Entrez Gene ID: (Human) 8556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support