Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LTVPESHRKP LKRSSSETIL SSLAGSDIVK GHEETCIRRA KAVKQRGISL LHEVDTQYSA LKVKYEELLK KCQEEQDSLS HKAVQTSRAA AKDLTGVNAQ SEPVASGWE
Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cerebellar degeneration-related protein 2; cerebellar degeneration-related protein 2, 62kDa; Major Yo paraneoplastic antigen; Paraneoplastic cerebellar degeneration-associated antigen; Yo paraneoplastic antigen
Gene Aliases: CDR2; CDR62; PCD17; Yo
UniProt ID: (Human) Q01850
Entrez Gene ID: (Human) 1039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support