Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VWGSPYSPPR EDWSVYPGPS STMGTVPVND VTSSPAAFCS TDYSNLGPVG GGTSGSSLPG QAGGSLVPTD AGAAKASSPS RSRHSPYAWM RKTVQVTGKT
This gene encodes a member of a small subfamily of homeobox containing transcription factors. The encoded protein may regulate homeobox gene expression during anteroposterior patterning and hematopoiesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caudal type homeo box 4; caudal type homeobox transcription factor 4; Caudal-type homeobox protein 4; Homeobox protein CDX-4
Gene Aliases: Cdx-3; Cdx-4; Cdx3; CDX4; RGD1561529
UniProt ID: (Human) O14627, (Mouse) Q07424
Entrez Gene ID: (Human) 1046, (Mouse) 12592, (Rat) 302400
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support