Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A431, HeLa.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asterless; CC-CKR-8; CCR-8; centrosomal protein 152kDa; Centrosomal protein of 152 kDa; Cep152; CKRL1; CMKBR8; CY6; microcephaly, primary autosomal recessive 4
Gene Aliases: CEP152; KIAA0912; MCPH4; MCPH9; SCKL5
UniProt ID: (Human) O94986
Entrez Gene ID: (Human) 22995
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support