Plays a critical role in cell cycle progression.
Protein Aliases: Proline-rich protein 11; transcription repressor of MHCII
Gene Aliases: PRR11
UniProt ID: (Human) Q96HE9
Entrez Gene ID: (Human) 55771