Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Blocks Notch protein; Botch; Cation transport regulator-like protein 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; Glutathione-specific gamma-glutamylcyclotransferase 1
Gene Aliases: BOTCH; CHAC1
UniProt ID: (Human) Q9BUX1
Entrez Gene ID: (Human) 79094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support