Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EEERLIRHLF QEKGYNKELR PVAHKEESVD VALALTLSNL ISLKEVEETL TTNVWIEHGW TDNRLKWNAE EFGNISVLRL PPDMVWLPEI VLENNNDGSF QISYSCN
The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylcholine receptor; Acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); cholinergic receptor; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide
Gene Aliases: ACHRD; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; FCCMS; SCCMS
UniProt ID: (Human) Q07001
Entrez Gene ID: (Human) 1144
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support