Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Control: K562 cell lysates, human liver tissue lysate, human skeletal muscle tissue lysate, HepG2, human fetal skeletal muscle tissue, human liver tissue, SH-SY5Y.
Subcellular Location: Cytoplasm, Membrane.
Chloride intracellular channel 2 is a member of the p64 family, a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. CLIon Channel 2 encodes a protein that is detected in fetal liver and adult skeletal muscle tissue. CLIon Channel2 maps to the candidate region on chromosome X for incontinentia pigmenti.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chloride intracellular channel protein 2; XAP121
Gene Aliases: CLIC2; CLIC2b; MRXS32; XAP121
UniProt ID: (Human) O15247
Entrez Gene ID: (Human) 1193
Molecular Function: ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support