Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSAEVKVTG QNQEQFLLLA KSAKGAALAT LIHQVLEAPG VYVFGELLDM PNVRELAESD FASTFRLLTV FAYGTYADYL AEARNLPPLT EAQKNKLRHL SVVTLAAKVK CIPYAVLLEA LALRNVRQLE DLVIEAVYAD VLRGSLDQRN QRLEVDYSIG RDIQRQDLSA IARTLQEWCV GCEVVLSGIE EQVSRANQHK EQQLGLKQQI ESEVANLKKT IKVTTAAAAA ATSQDPEQHL TELREPAPGT NQRQPSKKAS KGKGLRGSAK IWSKSN (1-275 aa encoded by B C011789)
The COP9 signalosome (CSN) complex is involved in several different developmental and cellular processes. The complex is made up of several widely expressed proteins: CSN1 (COPS1), CSN2 (COPS2), CSN3 (COPS3), CSN4 (COPS4), CSN5 (COPS5), CSN6 (COP6), CSN7a (COPS7, COPS7a) or CSN7b (COP7b) and CSN8 (COP8). The CSN complex acts as a regulator for the ubiquitin conjugation pathway by mediating the deneddylation of the SCF-type E3 ligase complexes, which leads to a decrease in ubiquitin ligase activity of SCF-complexes. It is also involved in the phosphorylation of p53, c-Jun, I B-alpha and IRF-8, as well as CSN-dependent phosphorylation of p53, and c-Jun protects and promotes degradation by the Ubl system. CSN7 is phosphorylated by CK2 and is composed of two subunits; a and b. CSN7a contains a PCI (Proteasome CSN9 initiation factor 3) region, as well as a coiled-coil region and is predicted to interact with CSN2, CSN3, CSN4, CSN5, CSN6, CSN8, and GPS1. CSN7b contains only a PCI region and is predicted to interact with INT6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COP9 (constitutive photomorphogenic) homolog, subunit 7a; COP9 (constitutive photomorphogenic), subunit 7a; COP9 7a; COP9 complex S7a; COP9 complex subunit 7a; COP9 constitutive photomorphogenic homolog subunit 7A; COPS7a; dermal papilla-derived protein 10; JAB1-containing signalosome subunit 7a; signalosome subunit 7a; unnamed protein product; ZH11
Gene Aliases: CSN7; CSN7A; D6Ertd35e; SGN7a
UniProt ID: (Human) Q9UBW8, (Mouse) Q9CZ04
Entrez Gene ID: (Human) 50813, (Mouse) 26894, (Rat) 312710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support