Product References
Brief Communication: Histological Assessment of Nonhuman Primate Brown Adipose Tissue Highlights the Importance of Sympathetic Innervation.
Journal of obesity
Williams AG,Long M,Kavanagh K
MA5-15078 was used in Immunohistochemistry to support the development of safe sympathetic nervous system stimulants to activate brown adipose tissue for obesity treatment.
Thu Feb 02 00:00:00 EST 2023
Brief Communication: Histological Assessment of Nonhuman Primate Brown Adipose Tissue Highlights the Importance of Sympathetic Innervation.
Journal of obesity
Williams AG,Long M,Kavanagh K
MA5-15078 was used in Immunohistochemistry to support the development of safe sympathetic nervous system stimulants to activate brown adipose tissue for obesity treatment.
Thu Feb 02 00:00:00 EST 2023
Brief Communication: Histological Assessment of Nonhuman Primate Brown Adipose Tissue Highlights the Importance of Sympathetic Innervation.
Journal of obesity
Williams AG,Long M,Kavanagh K
MA5-15078 was used in Immunohistochemistry to support the development of safe sympathetic nervous system stimulants to activate brown adipose tissue for obesity treatment.
Thu Feb 02 00:00:00 EST 2023
MiR-27b attenuates mitochondrial oxidative stress and inflammation in endothelial cells.
Redox biology
D'Onofrio N,Prattichizzo F,Martino E,Anastasio C,Mele L,La Grotta R,Sardu C,Ceriello A,Marfella R,Paolisso G,Balestrieri ML
MA5-15078 was used in Western Blotting to reveal for the first time that miR-27b could represent a possible target for future therapies aimed at improving endothelial health.
Thu Jun 01 00:00:00 EDT 2023
Cardiac disruption of SDHAF4-mediated mitochondrial complex II assembly promotes dilated cardiomyopathy.
Nature communications
Wang X,Zhang X,Cao K,Zeng M,Fu X,Zheng A,Zhang F,Gao F,Zou X,Li H,Li M,Lv W,Xu J,Long J,Zang W,Chen J,Gao F,Ding J,Liu J,Feng Z
MA5-15078 was used in Western Blotting to reveal a vital role for complex II assembly in the development of dilated cardiomyopathy and provide additional insights into therapeutic interventions for heart diseases.
Fri Jul 08 00:00:00 EDT 2022
Tick-borne encephalitis virus capsid protein induces translational shutoff as revealed by its structural-biological analysis.
The Journal of biological chemistry
Selinger M,Novotný R,Sýs J,Roby JA,Tykalová H,Ranjani GS,Vancová M,Jaklová K,Kaufman F,Bloom ME,Zdráhal Z,Grubhoffer L,Forwood JK,Hrabal R,Rumlová M,Štěrba J
MA5-15078 was used in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot to show the first capsid protein 3D structure of a member of the tick-borne flaviviruses group and show that by metabolic labeling experiments, TBEVC and other flaviviral capsid proteins are able to induce translational shutoff and decrease of 18S rRNA.
Tue Nov 01 00:00:00 EDT 2022
On the potential role of globins in brown adipose tissue: a novel conceptual model and studies in myoglobin knockout mice.
American journal of physiology. Endocrinology and metabolism
Blackburn ML,Wankhade UD,Ono-Moore KD,Chintapalli SV,Fox R,Rutkowsky JM,Willis BJ,Tolentino T,Lloyd KCK,Adams SH
MA5-15078 was used in Western Blotting to suggest a role for myoglobin as an oxygen/nitric oxide-sensor that regulates cellular metabolic and signaling pathways.
Thu Jul 01 00:00:00 EDT 2021
Abnormal expression of lncRNA UCA1 disturbed cell apoptosis through mediating mitochondrial dynamics in PDAC.
Neoplasma
Teng B,Feng T,Li W,Wang Z
MA5-15078 was used in Western Blot to report that UCA1-driven change in mitochondrial dynamics induces mitochondrial apoptotic pathway in PDAC.
Mon Mar 01 00:00:00 EST 2021
Interspecies Differences in Proteome Turnover Kinetics Are Correlated With Life Spans and Energetic Demands.
Molecular & cellular proteomics : MCP
Swovick K,Firsanov D,Welle KA,Hryhorenko JR,Wise JP,George C,Sformo TL,Seluanov A,Gorbunova V,Ghaemmaghami S
Published figure using COX4 monoclonal antibody (Product # MA5-15078) in Western Blot
Fri Mar 11 00:00:00 EST 2022
Interspecies Differences in Proteome Turnover Kinetics Are Correlated With Life Spans and Energetic Demands.
Molecular & cellular proteomics : MCP
Swovick K,Firsanov D,Welle KA,Hryhorenko JR,Wise JP,George C,Sformo TL,Seluanov A,Gorbunova V,Ghaemmaghami S
Published figure using COX4 monoclonal antibody (Product # MA5-15078) in Western Blot
Fri Mar 11 00:00:00 EST 2022
Colorectal Cancer Apoptosis Induced by Dietary δ-Valerobetaine Involves PINK1/Parkin Dependent-Mitophagy and SIRT3.
International journal of molecular sciences
D'Onofrio N,Martino E,Mele L,Colloca A,Maione M,Cautela D,Castaldo D,Balestrieri ML
MA5-15078 was used in Western Blotting to support a critical role of PINK1/Parkin-mediated mitophagy in mitochondrial dysfunction and apoptosis induced by δVB in SW480 and SW620 colon cancer cells.
Thu Jul 29 00:00:00 EDT 2021
PARK14 PLA2G6 mutants are defective in preventing rotenone-induced mitochondrial dysfunction, ROS generation and activation of mitochondrial apoptotic pathway.
Oncotarget
Chiu CC,Yeh TH,Lu CS,Huang YC,Cheng YC,Huang YZ,Weng YH,Liu YC,Lai SC,Chen YL,Chen YJ,Chen CL,Chen HY,Lin YW,Wang HL
Published figure using COX4 monoclonal antibody (Product # MA5-15078) in Western Blot
Tue Oct 03 00:00:00 EDT 2017
A nuclear role for the respiratory enzyme CLK-1 in regulating mitochondrial stress responses and longevity.
Nature cell biology
Monaghan RM,Barnes RG,Fisher K,Andreou T,Rooney N,Poulin GB,Whitmarsh AJ
MA5-15078 was used in western blot to study the role of nuclear CLK-1 in regulating mitochondrial stress responses and aging.
Mon Jun 01 00:00:00 EDT 2015
A nuclear role for the respiratory enzyme CLK-1 in regulating mitochondrial stress responses and longevity.
Nature cell biology
Monaghan RM,Barnes RG,Fisher K,Andreou T,Rooney N,Poulin GB,Whitmarsh AJ
MA5-15078 was used in western blot to study the role of nuclear CLK-1 in regulating mitochondrial stress responses and aging.
Mon Jun 01 00:00:00 EDT 2015
3,5-Diiodo-L-thyronine activates brown adipose tissue thermogenesis in hypothyroid rats.
PloS one
Lombardi A,Senese R,De Matteis R,Busiello RA,Cioffi F,Goglia F,Lanni A
Published figure using COX4 monoclonal antibody (Product # MA5-15078) in Immunohistochemistry
Fri Jan 15 00:00:00 EST 2016