Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PVHAQPPGTP LSQAAASIDN RVPTPSSVAS AETNSQQPGP DVPVLEMKTE TQAEDTEPDP GESKGEPRSE MMEEDLQGAS QVKEETDIAE QKSEPMEVDE
CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CREB-binding protein; Histone lysine acetyltransferase CREBBP; Protein lactyltransferas CREBBP; Protein-lysine acetyltransferase CREBBP
Gene Aliases: CBP; CREBBP; KAT3A; RSTS
UniProt ID: (Human) Q92793
Entrez Gene ID: (Human) 1387
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support