Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TTSLFRDSPAG DHDYALPVGK QKQDLLEEDD SAGGVCLHVD KDKVSVEFCS ACARKASSSL
Highest antigen sequence identity to the following orthologs - mouse 90%, rat 95%.
Strongly activates transcription when bound to HCFC1. Suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1-dependent manner. Also suppresses the HCFC1-dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell. Able to down-regulate expression of some cellular genes in CREBZF-expressing cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support