Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MMAYMNPGPH YSVNALALSG PSVDLMHQAV PYPSAPRKQR RERTTFTRSQ LEELEALFAK TQYPDVYARE EVALKINLPE SRVQVWFKNR RAKCRQQRQQ QKQQQQPPGG QAKARPAKRK AGTSPRPSTD VCPDPLGISD SYSPPLPGPS GSPTTAVATV SIWSPASESP LPEAQRAGLV ASGPSLTSAP YAMTYAPASA FCSSPSAYGS PSSYFSGLDP YLSPMVPQLG GPALSPLSGP SVGPSLAQSP TSLSGQSYGA YSPVDSLEFK DPTGTWKFTY NPMDPLDYKD QSAWKFQIL
The cone-rod homeobox-containing gene (CRX) encodes a transcription factor that coordinates the expression of several photoreceptor genes in the developing retina, including opsin and rhodopsin. Specifically, CRX binds the OTX motif (TAATCC/A) upstream from photoreceptor genes. The CRX gene is also expressed in the pinealocytes of the pineal gland and may regulate pineal circadian activity by controling the expression of melatonin synthesis genes. Furthermore, CRX(-) mice exhibit disruption of circadian rhythms. The human CRX gene maps to chromosome 19q13.3 within the region of the cone-rod dystrophy-2 locus (CORD2). Mutations in the CRX gene are implicated in the visual pathologies of CORD, Leber congenital amaurosis (LCA) and retinitis pigmentosa (RP). All characterized CRX gene mutations produce disease in heterozygotes although there is no known correlation between the pathologic phenotype and genetic mutation. Missense mutations of the CRX gene affect the homeobox domain, whereas frameshift mutations affect the OTX domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cone rod homeobox; Cone-rod homeobox protein; CORD 2; LCA 7; orthodenticle homeobox 3; OTX; OTX 3
Gene Aliases: CORD2; CRD; CRX; LCA7; OTX3
UniProt ID: (Human) O43186
Entrez Gene ID: (Human) 1406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support