Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
Positive control: Highly expressed in heart and throughout the brain, with similar levels in cortex and hypothalamus, and much higher levels in hippocampus, dentate gyrus and the habenular nucleus of the thalamus. Detected at similar levels in heart atrium and ventricle. Detected in aorta, cardiac and smooth muscle.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium-activated neutral proteinase 1; calpain 1, (mu/I) large subunit; Calpain mu-type; calpain, large polypeptide L1; Calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP 1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; micromolar calcium-activated neutral protease 1; Micromolar-calpain; mu-calpain; mucalpain; muCANP
Gene Aliases: CANP; CANP 1; CANP1; CANPL1; Capa-1; Capa1; CAPN1; Cls1; mu-calpin; muCANP; muCL; PIG30; SPG76
UniProt ID: (Bovine) Q27970, (Human) P07384, (Mouse) O35350, (Rat) P97571
Entrez Gene ID: (Bovine) 281661, (Human) 823, (Mouse) 12333, (Rat) 29153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support