Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
In Western blots, no cross-reactivity with recombinant human CA1, 4, or CA6-14 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase. CA5B belongs a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. This protein is localized in the mitochondria and shows the highest sequence similarity to the other mitochondrial CA, CA VA. The protein has a wider tissue distribution than CA VA, which is restricted to the liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CA-VA; CA-VB; Carbonate dehydratase VA; Carbonate dehydratase VB; Carbonic anhydrase 5A, mitochondrial; Carbonic anhydrase 5B, mitochondrial; carbonic anhydrase V, mitochondrial; Carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; Carbonic anhydrase VB; carbonic anhydrase VB, mitochondrial; carbonic dehydratase
Gene Aliases: CA-VB; CA5; CA5A; CA5AD; CA5B; CAV; CAVA; GS1-21A4.1
UniProt ID: (Human) P35218, (Human) Q9Y2D0
Entrez Gene ID: (Human) 763, (Human) 11238
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support