Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to Chikungunya virus and does not cross-react with other Alphaviruses Eastern Equine Encephalitis, Western Equine Encephalitis, Venezuelan Equine Encephalitis or other Arboviruses.
Purity is >90%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHIKV
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support