Invitrogen
MA1-7337 is specific for MOMP, and recognizes 15 serovars of Chlamydia trachomatis.
MA1-7337 has been successfully used in ELISA, immunofluorescence and immunohistochemical applications.
The MA1-7337 immunogen is a C. trachomatis preparation.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.