Search Thermo Fisher Scientific
PA1-27223 detects C. trachomatis from bacteria samples.
PA1-27223 has been successfully used in immunofluorescence and ELISA procedures.
The PA1-27223 immunogen is L2 and other serovar groups.
Store at 4ºC short term. For extended storage, aliquot and store at -20ºC, avoiding freeze/thaw cycles.
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.