Product References
Bevacizumab plus octreotide and metronomic capecitabine in patients with metastatic well-to-moderately differentiated neuroendocrine tumors: the XELBEVOCT study.
BMC cancer
Berruti A,Fazio N,Ferrero A,Brizzi MP,Volante M,Nobili E,Tozzi L,Bodei L,Torta M,D'Avolio A,Priola AM,Birocco N,Amoroso V,Biasco G,Papotti M,Dogliotti L
MA5-13093 was used in immunohistochemistry to study the performance of the XELBEVOCT therapeutic regimen in patients with well-to-moderately differentiated neuroendocrine tumors
Fri Mar 14 00:00:00 EDT 2014
Tumor staging but not grading is associated with adverse clinical outcome in neuroendocrine tumors of the appendix: a retrospective clinical pathologic analysis of 138 cases.
The American journal of surgical pathology
Volante M,Daniele L,Asioli S,Cassoni P,Comino A,Coverlizza S,De Giuli P,Fava C,Manini C,Berruti A,Papotti M
MA5-13093 was used in immunohistochemistry to study the association of tumor staging with clinical outcome in neuroendocrine tumors of the appendix
Mon Apr 01 00:00:00 EDT 2013
Human ASH-1 promotes neuroendocrine differentiation in androgen deprivation conditions and interferes with androgen responsiveness in prostate cancer cells.
The Prostate
Rapa I,Volante M,Migliore C,Farsetti A,Berruti A,Vittorio Scagliotti G,Giordano S,Papotti M
MA5-13093 was used in immunohistochemistry to study the role of the human ASH-1 transcription factor in the development of the prostate cancer cell neuroendocrine phenotype
Thu Aug 01 00:00:00 EDT 2013
Human papillomavirus-associated adenocarcinoma of the base of the tongue.
Human pathology
Hanna J,Reimann JD,Haddad RI,Krane JF
MA5-13093 was used in immunohistochemistry to report on two cases of adenocarcinoma of the base of the tongue associated with HPV infection
Thu Aug 01 00:00:00 EDT 2013
Gastric histopathological findings and ghrelin expression in morbid obesity.
Turk patoloji dergisi
Gündoğan M,Çalli Demırkan N,Tekın K,Aybek H
MA5-13093 was used in immunohistochemistry to study the histopathology of ghrelin-expressing cells in the stomach of morbidly obese patients
Tue Feb 04 00:00:00 EST 2014
Airway basal cell vascular endothelial growth factor-mediated cross-talk regulates endothelial cell-dependent growth support of human airway basal cells.
Cellular and molecular life sciences : CMLS
Curradi G,Walters MS,Ding BS,Rafii S,Hackett NR,Crystal RG
MA5-13093 was used in immunohistochemistry to study the expression VEGF isoforms by airway basal cells and the mechanism by which they regulate endothelial activation
Sun Jul 01 00:00:00 EDT 2012
HPV-associated neuroendocrine carcinoma of the oropharynx: a rare new entity with potentially aggressive clinical behavior.
The American journal of surgical pathology
Kraft S,Faquin WC,Krane JF
MA5-13093 was used in immunohistochemistry to report on 8 cases of HPV- and tobacco-associated neuroendocrine carcinoma of the oropharynx
Thu Mar 01 00:00:00 EST 2012
EGF promotes neuroendocrine-like differentiation of prostate cancer cells in the presence of LY294002 through increased ErbB2 expression independent of the phosphatidylinositol 3-kinase-AKT pathway.
Carcinogenesis
Cortés MA,Cariaga-Martinez AE,Lobo MV,Martín Orozco RM,Motiño O,Rodríguez-Ubreva FJ,Angulo J,López-Ruiz P,Colás B
MA5-13093 was used in immunocytochemistry and immunohistochemistry to study the mechanism by which EGF promotes neuroendocrine-like differention of prostate cancer cells in the presence of a PI3 kinase inhibitor
Fri Jun 01 00:00:00 EDT 2012
Human achaete-scute homolog-1 expression in neuroendocrine breast carcinoma.
Virchows Archiv : an international journal of pathology
Righi L,Rapa I,Votta A,Papotti M,Sapino A
MA5-13093 was used in immunohistochemistry to study the expression of the hASH-1 transcription factor in neuroendocrine breast carcinoma
Sun Apr 01 00:00:00 EDT 2012
Extraventricular neurocytomas: a morphological and histogenetic consideration. A study of six cases.
Pathology
Agarwal S,Sharma MC,Sarkar C,Suri V,Jain A,Sharma MS,Ailawadhi P,Garg A,Mallick S
MA5-13093 was used in immunohistochemistry to study the morphological and histological features of neurocytomas in extraventricular locations
Wed Jun 01 00:00:00 EDT 2011
Detection of bone marrow metastases of neuroblastoma with immunohistochemical staining of CD56, chromogranin A, and synaptophysin.
Applied immunohistochemistry & molecular morphology : AIMM
Park SJ,Park CJ,Kim S,Jang S,Chi HS,Kim MJ,Im HJ,Seo JJ
MA5-13093 was used in immunohistochemistry to detect the bone marrow metastases of neuroblastoma by immunohistochemical staining of specific proteins
Thu Jul 01 00:00:00 EDT 2010
Pulmonary meningothelial-like nodules: new insights into a common but poorly understood entity.
The American journal of surgical pathology
Mukhopadhyay S,El-Zammar OA,Katzenstein AL
MA5-13093 was used in immunohistochemistry to study pulmonary meningothelial-like nodules
Wed Apr 01 00:00:00 EDT 2009
Extra-adrenal composite paraganglioma with ganglioneuroma component presenting as a pancreatic mass.
Endocrine pathology
Inzani F,Rindi G,Tamborrino E,Cobelli R,Bordi C
MA5-13093 was used in immunohistochemistry to report on a case of extra-adrenal composite paraganglioma with ganglioneuroma component
Tue Nov 17 00:00:00 EST 2009
Chromogranin peptides in amyotrophic lateral sclerosis.
Regulatory peptides
Schrott-Fischer A,Bitsche M,Humpel C,Walcher C,Maier H,Jellinger K,Rabl W,Glueckert R,Marksteiner J
MA5-13093 was used in western blot to investigate the role of chromogranin peptides in amyotrophic lateral sclerosis
Thu Jan 08 00:00:00 EST 2009
Basaloid adenocarcinoma. A new variant of pulmonary adenocarcinoma.
Virchows Archiv : an international journal of pathology
Marci V,Volante M,Cappia S,Righi L,Novello C,Scagliotti GV,Brambilla E,Papotti M
MA5-13093 was used in immunohistochemistry to report a clinical case of basaloid adenocarcinoma
Sat Sep 01 00:00:00 EDT 2007
Primary neuroendocrine carcinoma of the breast: a case report.
Tumori
Yaren A,Kelten C,Akbulut M,Teke Z,Duzcan E,Erdem E
MA5-13093 was used in immunohistochemistry to report on a case of primary neuroendocrine carcinoma of the breast
Tue Dec 11 00:00:00 EST 2007
Laparoscopic resection of urinary bladder paraganglioma: a case report.
Surgical laparoscopy, endoscopy & percutaneous techniques
Dilbaz B,Bayoglu Y,Oral S,Cavusoglu D,Uluoglu O,Dilbaz S
MA5-13093 was used in immunohistochemistry to report on a case of aparoscopic resection of urinary bladder paraganglioma
Wed Feb 01 00:00:00 EST 2006
Two functional epitopes of pigment epithelial-derived factor block angiogenesis and induce differentiation in prostate cancer.
Cancer research
Filleur S,Volz K,Nelius T,Mirochnik Y,Huang H,Zaichuk TA,Aymerich MS,Becerra SP,Yap R,Veliceasa D,Shroff EH,Volpert OV
MA5-13093 was used in western blot to investigate the role of two functional epitopes of pigment epithelial-derived factor in prostate cancer
Wed Jun 15 00:00:00 EDT 2005
Ovarian ependymoma.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Erdoğan G,Ozel E,Peştereli HE,Salar Z,Tirak B,Karaveli S
MA5-13093 was used in immunohistochemistry to report on a case of anaplastic ependymoma.
Fri Apr 01 00:00:00 EST 2005
Pulmonary large cell carcinoma with rhabdoid phenotype.
Annals of diagnostic pathology
Yilmazbayhan D,Ates LE,Dilege S,Gulluoglu M,Tanju S,Kalayci G
MA5-13093 was used in immunohistochemistry The report on a patient with pulmonary large cell carcinoma with rhabdoid phenotype
Mon Aug 01 00:00:00 EDT 2005
CDX-2 homeobox gene product expression in neuroendocrine tumors: its role as a marker of intestinal neuroendocrine tumors.
The American journal of surgical pathology
Barbareschi M,Roldo C,Zamboni G,Capelli P,Cavazza A,Macri E,Cangi MG,Chilosi M,Doglioni C
MA5-13093 was used in immunohistochemistry to study the expression of the CDX-2 homeobox gene product in neuroendocrine tumors
Wed Sep 01 00:00:00 EDT 2004
Altered distribution of metaplastic Paneth, gastrin and pancreatic acinar cells in atrophic gastritic mucosa with endocrine cell lesions.
The Tohoku journal of experimental medicine
Deveci MS,Deveci G
MA5-13093 was used in immunohistochemistry to study the altered distribution of metaplastic cell types in atrophic gastritic mucosa with endocrine cell lesions
Thu Jan 01 00:00:00 EST 2004
Concurrent lymph node metastases of medullary and papillary thyroid carcinoma in a case with RET oncogene germline mutation.
Endocrine pathology
Papi G,Corrado S,Pomponi MG,Carapezzi C,Cesinaro A,LiVolsi VA
MA5-13093 was used in immunohistochemistry to report on a case of pituitary cancer with lymph node metastasis in a patient with a RET ongogene germline mutation
Tue Jun 29 00:00:00 EDT 2004
Heparin-binding epidermal growth factor-like growth factor stimulates androgen-independent prostate tumor growth and antagonizes androgen receptor function.
Endocrinology
Adam RM,Kim J,Lin J,Orsola A,Zhuang L,Rice DC,Freeman MR
MA5-13093 was used in western blot to investigate the influence of heparin-binding epidermal growth factor-like growth factor on prostate tumor growth
Sun Dec 01 00:00:00 EST 2002
Classification of low-grade neuroendocrine tumors of midgut and unknown origin.
Human pathology
Van Eeden S,Quaedvlieg PF,Taal BG,Offerhaus GJ,Lamers CB,Van Velthuysen ML
MA5-13093 was used in immunohistochemistry to study whether refining the grading of low-grade neuroendocrine tumors of the midgut and unknown origin is of prognostic/predictive value
Fri Nov 01 00:00:00 EST 2002