Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: VVGGEDARPH SWPWQISLQY LKNDTWRHTC GGTLIASNFV LTAAHCISNT RTYRVAVGKN NLEVEDEEGS LFVGVDTIHV HKRWNALLLR NDIALIKLAE HVELSDTIQV ACLPEKDSLL P
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chymotrypsin-C
Gene Aliases: 1810044E12Rik; Ctrc
UniProt ID: (Mouse) Q3SYP2
Entrez Gene ID: (Mouse) 76701
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support