Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-40065 detects Collagen II from mouse, rat, bovine, chicken, and human samples.
MA1-40065 has been successfully used in immunohistochemistry (frozen), ELISA, funtional assays and immunohistochemistry (paraffin) applications.
The MA1-40065 immunogen is mouse Collagen II.
Endotoxin <24 EU/mg.
COL2A1 is the alpha-1 chain of type II collagen, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia, Stickler syndrome type I, and spondyloepimetaphyseal dysplasia Strudwick type. In addition, defects in processing chondrocalcin, a calcium binding protein that is the C-propeptide of this collagen molecule, are also associated with chondrodysplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha 1 type II procollagen; alpha 1 type IIA collagen; alpha collagen type II; Alpha-1 type II collagen; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; CGA; chondrocalcin; Collagen 2; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen type II (Col2A1) gene, enhancer region; collagen, type II, alpha 1; collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphyseal dysplasia, congenital); disproportionate micromelia; MGC131516; pro-alpha 1(II); procollagen alpha 1(II); Procollagen II alpha 1; procollagen, type II, alpha 1; type II collagen; unnamed protein product
Gene Aliases: ANFH; AOM; CB11; CG2A1A; COL11A3; Col2; Col2a; Col2a-1; COL2A1; COLLII; Del1; Dmm; Lpk; M100856; Rgsc856; SEDC; STL1
UniProt ID: (Human) P02458, (Bovine) P02459, (Rat) Q78DY3, (Mouse) P28481
Entrez Gene ID: (Human) 1280, (Chicken) 395069, (Bovine) 407142, (Rat) 25412, (Mouse) 12824
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support