Invitrogen
Mouse anti Cryptosporidium antibody, clone BEL 0126 recognizes a membrane antigen expressed by the oocysts of Cryptosporidium sp., an obligate enteric coccidian parasite of the phylum Apicomplexa that infects the gastrointestinal tract.
Protein Aliases: Cryptosporidiosis