Search Thermo Fisher Scientific
PA1-24255 detects Cyclin G2 in Human samples.
PA1-24255 has been successfully used in Western Blot procedures.
The PA1-24255 immunogen is a synthetic peptide corresponding to a region of human CCNG2.
Cyclin G2 may play a role in growth regulation and in negative regulation of cell cycle progression.