Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Especially useful in discrimination of morphologically altered "round-headed" spermatozoa. Polypeptides reacting: Multiple band proteins with MW 63, 69 and 74 kDa.
Possible architectural role during spermatogenesis. May be involved in spermatid differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cylicin I; cylicin, basic protein of sperm head cytoskeleton 1; Cylicin-1; Multiple-band polypeptide I
Gene Aliases: CYL; CYL1; CYLC1
UniProt ID: (Bovine) P35662
Entrez Gene ID: (Bovine) 281737
Molecular Function:
structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support