Product References
The Hippo kinases LATS1 and 2 control human breast cell fate via crosstalk with ERα.
Nature
Britschgi A,Duss S,Kim S,Couto JP,Brinkhaus H,Koren S,De Silva D,Mertz KD,Kaup D,Varga Z,Voshol H,Vissieres A,Leroy C,Roloff T,Stadler MB,Scheel CH,Miraglia LJ,Orth AP,Bonamy GM,Reddy VA,Bentires-Alj M
MA112589 was used in immunocytochemistry and western blot to define the interaction between LATS1/2 and the estrogen receptor signaling in breast cancer initiation.
Thu Jan 26 00:00:00 EST 2017
Vitamin D and androgen receptor-targeted therapy for triple-negative breast cancer.
Breast cancer research and treatment
Thakkar A,Wang B,Picon-Ruiz M,Buchwald P,Ince TA
MA1-12589 was used in western blot to characterize androgen receptor-targeted and vitamin D therapy for triple-negative breast cancer
Sun May 01 00:00:00 EDT 2016
FAM83H and casein kinase I regulate the organization of the keratin cytoskeleton and formation of desmosomes.
Scientific reports
Kuga T,Sasaki M,Mikami T,Miake Y,Adachi J,Shimizu M,Saito Y,Koura M,Takeda Y,Matsuda J,Tomonaga T,Nakayama Y
MA112589 was used in immunocytochemistry and western blot to examine the effects of the FAM83H mutation on the keratin cytoskeleton and desmosomes in ameloblasts
Wed May 25 00:00:00 EDT 2016
Casein kinase 1 is recruited to nuclear speckles by FAM83H and SON.
Scientific reports
Kuga T,Kume H,Adachi J,Kawasaki N,Shimizu M,Hoshino I,Matsubara H,Saito Y,Nakayama Y,Tomonaga T
MA112589 was used in immunocytochemistry, immunohistochemistry - paraffin section, and western blot to study factors that influence the localization of casein kinase 1
Thu Sep 29 00:00:00 EDT 2016
Modelling breast cancer requires identification and correction of a critical cell lineage-dependent transduction bias.
Nature communications
Hines WC,Yaswen P,Bissell MJ
MA1-12589 was used in immunocytochemistry to study and identify critical cell lineage-dependent transduction bias to model breast cancer properly
Tue Apr 21 00:00:00 EDT 2015
Alterations of canalicular ATP-binding cassette transporter expression in drug-induced liver injury.
Digestion
Zollner G,Thueringer A,Lackner C,Fickert P,Trauner M
MA1-12589 was used in immunohistochemistry - paraffin section to evaluate the role of canalicular ATP-binding cassette transporter expression in drug-induced liver injury
Wed Jun 24 00:00:00 EDT 2015
Multiple oncocytic cystadenoma with intraluminal crystalloids in parotid gland: case report.
Medicine
Başak K,Kroğlu K
MA1-12589 was used in immunohistochemistry to describe a tumor from a patient with oncocytic cystadenoma
Mon Dec 01 00:00:00 EST 2014
Glucose and SIRT2 reciprocally mediate the regulation of keratin 8 by lysine acetylation.
The Journal of cell biology
Snider NT,Leonard JM,Kwan R,Griggs NW,Rui L,Omary MB
MA1-12589 was used in immunoprecipitation to study the opposing effects of hyperglycemia and SIRT2 on keratin-8 lysine acetylation
Mon Feb 04 00:00:00 EST 2013
Malignant transformation of adenomyoepithelioma of the breast by a monophasic population: a report of two cases and review of literature.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Marian C,Boila A,Soanca D,Malau M,Podeanu DM,Resetkova E,Stolnicu S
MA1-12589 was used in immunohistochemistry to report on two cases of malignant transformation of adenomyoepithelioma of the breast by a monophasic population
Mon Apr 01 00:00:00 EDT 2013
A small molecule inhibitor of SRC family kinases promotes simple epithelial differentiation of human pluripotent stem cells.
PloS one
Lian X,Selekman J,Bao X,Hsiao C,Zhu K,Palecek SP
MA1-12589 was used in flow cytometry, immunocytochemistry, and western blot to describe conditions that promote development of hPSC-derived simple epithelial cells
Tue Sep 17 00:00:00 EDT 2013
Nuclear shape changes are induced by knockdown of the SWI/SNF ATPase BRG1 and are independent of cytoskeletal connections.
PloS one
Imbalzano KM,Cohet N,Wu Q,Underwood JM,Imbalzano AN,Nickerson JA
MA1-12589 was used in immunocytochemistry to investigate the role of SWI/SNF ATPase BRG1 in nuclear morphology
Tue Aug 20 00:00:00 EDT 2013
Expression of KRT7 and WT1 differentiates precursor lesions of Wilms' tumours from those of papillary renal cell tumours and mucinous tubular and spindle cell carcinomas.
Virchows Archiv : an international journal of pathology
Szponar A,Kovacs G
MA1-12589 was used in immunohistochemistry to study the value of KRT7 and WT1 expression profiles in differentiating Wilms' tumour, papillary renal cell tumour and mucinous tubular and spindle cell carcinoma precursor lesions
Sun Apr 01 00:00:00 EDT 2012
Cytoskeletal keratin glycosylation protects epithelial tissue from injury.
Nature cell biology
Ku NO,Toivola DM,Strnad P,Omary MB
MA1-12589 was used in western blot to study the role of cytoskeletal keratin glycosylation in protecting epithelial tissue from injury
Wed Sep 01 00:00:00 EDT 2010
Endometrial profile of tamoxifen and low-dose estradiol combination therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research
Wood CE,Kaplan JR,Fontenot MB,Williams JK,Cline JM
MA1-12589 was used in immunohistochemistry to detect effects of estradiol and tamoxifen on endometrial health
Mon Feb 01 00:00:00 EST 2010
Antagonistic roles of Notch and p63 in controlling mammary epithelial cell fates.
Cell death and differentiation
Yalcin-Ozuysal O,Fiche M,Guitierrez M,Wagner KU,Raffoul W,Brisken C
MA1-12589 was used in western blot to investigate the role of Notch and p63 in regulation of mammary epithelial cell fates
Fri Oct 01 00:00:00 EDT 2010
Assessment of the tumorigenesis and drug susceptibility of three new canine mammary tumor cell lines.
Research in veterinary science
Chang CY,Chiou PP,Chen WJ,Li YH,Yiu JC,Cheng YH,Chen SD,Lin CT,Lai YS
MA1-12589 was used in western blot to study the tumorigenesis and drug susceptibility of three new canine mammary tumor cell lines
Thu Apr 01 00:00:00 EDT 2010
Giant clear cell hidradenoma of the knee.
Journal of cutaneous pathology
Yu G,Goodloe S,D'Angelis CA,McGrath BE,Chen F
MA1-12589 was used in immunohistochemistry to report on a case of giant clear cell hidradenoma of the knee
Wed Sep 01 00:00:00 EDT 2010
Imaging and analysis of 3D tumor spheroids enriched for a cancer stem cell phenotype.
Journal of biomolecular screening
Robertson FM,Ogasawara MA,Ye Z,Chu K,Pickei R,Debeb BG,Woodward WA,Hittelman WN,Cristofanilli M,Barsky SH
MA1-12589 was used in immunohistochemistry to study 3D tumor spheroids enriched for a cancer stem cell phenotype
Sun Aug 01 00:00:00 EDT 2010
Directed differentiation of human embryonic stem cells to epidermal progenitors.
Methods in molecular biology (Clifton, N.J.)
Metallo CM,Ji L,de Pablo JJ,Palecek SP
MA1-12589 was used in flow cytometry to discus a method for the differentiation of human embryonic stem cells into epidermal progenitors
Fri Jan 29 00:00:00 EST 2010
Long-term cultures of bone marrow-derived human mesenchymal stem cells frequently undergo spontaneous malignant transformation.
Cancer research
Røsland GV,Svendsen A,Torsvik A,Sobala E,McCormack E,Immervoll H,Mysliwietz J,Tonn JC,Goldbrunner R,Lønning PE,Bjerkvig R,Schichor C
MA1-12589 was used in immunohistochemistry to study malignant transformation in long-term cultures of bone marrow-derived human mesenchymal stem cells
Wed Jul 01 00:00:00 EDT 2009
Frzb, a secreted Wnt antagonist, decreases growth and invasiveness of fibrosarcoma cells associated with inhibition of Met signaling.
Cancer research
Guo Y,Xie J,Rubin E,Tang YX,Lin F,Zi X,Hoang BH
MA1-12589 was used in western blot to study the effect of the secreted Wnt antagonist Frzb on fibrosarcoma growth and metastasis
Thu May 01 00:00:00 EDT 2008
Monitoring of epithelial cell caspase activation via detection of durable keratin fragment formation.
The Journal of pathology
Tao GZ,Li DH,Zhou Q,Toivola DM,Strnad P,Sandesara N,Cheung RC,Hong A,Omary MB
MA1-12589 was used in ELISA to study epithelial cell caspase activation via detection of a durable keratin fragment
Sun Jun 01 00:00:00 EDT 2008
Analysis of keratin polypeptides 8 and 19 variants in inflammatory bowel disease.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association
Tao GZ,Strnad P,Zhou Q,Kamal A,Zhang L,Madani ND,Kugathasan S,Brant SR,Cho JH,Omary MB,Duerr RH
MA1-12589 was used in western blot to study whether variants of keratin 8 and keratin 19 are associated with inflammatory bowel disease
Sun Jul 01 00:00:00 EDT 2007
An oestrogen-dependent model of breast cancer created by transformation of normal human mammary epithelial cells.
Breast cancer research : BCR
Duss S,André S,Nicoulaz AL,Fiche M,Bonnefoi H,Brisken C,Iggo RD
MA1-12589 was used in immunocytochemistry and immunohistochemistry to develop an estrogen receptor-alpha-positive model of breast cancer using human mammary epithelial cells
Fri Sep 14 00:00:00 EDT 2007
Cellular and molecular mechanisms of abnormal calcification following ischemia-reperfusion injury in human liver transplantation.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc
Kalantari F,Miao D,Emadali A,Tzimas GN,Goltzman D,Vali H,Chevet E,Auguste P
MA1-12589 was used in immunohistochemistry to investigate the mechanism for liver calcification after transplantation
Thu Mar 01 00:00:00 EST 2007
Gene expression profiling of extracellular matrix as an effector of human hepatocyte phenotype in primary cell culture.
Toxicological sciences : an official journal of the Society of Toxicology
Page JL,Johnson MC,Olsavsky KM,Strom SC,Zarbl H,Omiecinski CJ
MA1-12589 was used in western blot to examine the biological responses imparted by Matrigel overlays on primary human hepatocyte cultures
Fri Jun 01 00:00:00 EDT 2007
Blocking Wnt/LRP5 signaling by a soluble receptor modulates the epithelial to mesenchymal transition and suppresses met and metalloproteinases in osteosarcoma Saos-2 cells.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society
Guo Y,Zi X,Koontz Z,Kim A,Xie J,Gorlick R,Holcombe RF,Hoang BH
MA1-12589 was used in western blot to investigate the role of LRP5 in osteosarcoma Saos-2 cells and their mesenchymal transition
Sun Jul 01 00:00:00 EDT 2007
An immunohistochemical profile of giant cell carcinoma of the larynx.
Auris, nasus, larynx
Gurbuz Y,Kose N,Aydin O,Ozturk M
MA1-12589 was used in immunohistochemistry to report on a case of giant cell carcinoma of the larynx
Sat Sep 01 00:00:00 EDT 2007
Mammary gland development in early pubertal female macaques.
Toxicologic pathology
Wood CE,Hester JM,Cline JM
MA1-12589 was used in immunohistochemistry to investigate the effect of estradiol on early ductal and lobular development
Mon Oct 01 00:00:00 EDT 2007
Identification of 14-3-3sigma as a contributor to drug resistance in human breast cancer cells using functional proteomic analysis.
Cancer research
Liu Y,Liu H,Han B,Zhang JT
MA1-12589 was used in western blot to study the role of 14-3-3sigma in drug resistance in human breast cancer cells
Wed Mar 15 00:00:00 EST 2006
Protein phosphatase-2A associates with and dephosphorylates keratin 8 after hyposmotic stress in a site- and cell-specific manner.
Journal of cell science
Tao GZ,Toivola DM,Zhou Q,Strnad P,Xu B,Michie SA,Omary MB
MA1-12589 was used in western blot to study the site-specific dephosphorylation of keratin 8 by protein phosphatase-2A and how this varies according to cell type
Sat Apr 01 00:00:00 EST 2006
Expression of Frzb/secreted Frizzled-related protein 3, a secreted Wnt antagonist, in human androgen-independent prostate cancer PC-3 cells suppresses tumor growth and cellular invasiveness.
Cancer research
Zi X,Guo Y,Simoneau AR,Hope C,Xie J,Holcombe RF,Hoang BH
MA1-12589 was used in western blot to investigate the suppressive effects of Frzb/sFRP3 expression on tumor growth and cellular invasiveness in PC-3 cells
Tue Nov 01 00:00:00 EST 2005
Keratin 8 and 18 hyperphosphorylation is a marker of progression of human liver disease.
Hepatology (Baltimore, Md.)
Toivola DM,Ku NO,Resurreccion EZ,Nelson DR,Wright TL,Omary MB
MA1-12589 was used in western blot to study keratin 8 and keratin 18 hyperphosphorylation as markers for progression of human liver disease
Sun Aug 01 00:00:00 EDT 2004
Cytokeratin expression in lichen amyloidosus and macular amyloidosis.
Journal of the European Academy of Dermatology and Venereology : JEADV
Apaydin R,Gürbüz Y,Bayramgürler D,Müezzinoglu B,Bilen N
MA1-12589 was used in immunohistochemistry to investigate the expression of cytokeratins in lichen and macular amyloidosis and their diagnostic value
Sat May 01 00:00:00 EDT 2004
Keratin mutation in transgenic mice predisposes to Fas but not TNF-induced apoptosis and massive liver injury.
Hepatology (Baltimore, Md.)
Ku NO,Soetikno RM,Omary MB
MA1-12589 was used in western blot to study the role of keratin mutations in the predisposition to Fas-induced apoptosis and massive liver injury
Thu May 01 00:00:00 EDT 2003
Keratin 8 mutations in patients with cryptogenic liver disease.
The New England journal of medicine
Ku NO,Gish R,Wright TL,Omary MB
MA1-12589 was used in western blot to study mutations of keratin 8 in cryptogenic liver disease patients
Thu May 24 00:00:00 EDT 2001