Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Western blot detects a band approximately 58 kDa (cytokeratin 5), 56 kD (cytokeratin 6) and 52 kD (cytokeratin 8).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa cytokeratin; 60-kDa keratin; alpha keratin; CK-5; CK-6A; CK-6D; CK-8; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; cytokeratin 8; Cytokeratin endo A; cytokeratin type II, component III; Cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-8; Cytokeratin-A; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; K6A; K8; kamp-keratin derived antimicrobial peptide; KDAMP; KERA; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6 alpha; keratin 6A, , type II; keratin 6A, type II; keratin 6B; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 8; keratin complex 2, gene 6a; keratin IV; keratin K6h; keratin, epidermal type II, K6A; Keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; Keratin-5; Keratin-6-alpha; Keratin-6A; keratin-6C; Keratin-8; krt5; mK6-alpha; tuftelin interacting protein 8; type II alpha-keratin IIA; type II alpha-keratin IIC; type II cytokeratin A (no.8); type-II keratin Kb12; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb8
Gene Aliases: 2310016L08Rik; 3300001P10Rik; AA960620; AL022697; AU019895; AW146334; CARD2; CK-6C; CK-6E; CK-8; CK5; CK6A; CK6C; CK6D; CK8; CYK8; CYKER; DDD; DDD1; EBS2; EndoA; K2C8; K5; K6A; K6C; K6D; K8; Ker2; KO; Krt-2.8; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; KRT2-8; KRT5; KRT5A; Krt6; KRT6A; Krt6b; KRT6C; KRT6D; KRT8; mK6[a]; MK6a; PC3; Sak57; Tfip8
UniProt ID: (Bovine) Q5XQN5, (Human) P13647, (Human) P02538, (Bovine) P05786, (Human) P05787, (Rat) Q6P6Q2, (Mouse) Q922U2, (Mouse) P50446, (Rat) Q10758, (Mouse) P11679
Entrez Gene ID: (Sheep) 100303605, (Chicken) 407779, (Rabbit) 100352658, (Bovine) 281268, (Dog) 100687760, (Goat) 102175207, (Human) 3852, (Dog) 486523, (Human) 3853, (Bovine) 614456, (Goat) 102174931, (Chicken) 408041, (Rabbit) 100348891, (Bovine) 281269, (Dog) 486513, (Chicken) 426896, (Goat) 102182762, (Sheep) 101110495, (Human) 3856, (Guinea pig) 100715346, (Rat) 369017, (Mouse) 110308, (Guinea pig) 100715629, (Mouse) 16687, (Guinea pig) 100736243, (Rat) 25626, (Chinese hamster) 100759488, (Mouse) 16691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support