Product References
Epithelial Fli1 deficiency drives systemic autoimmunity and fibrosis: Possible roles in scleroderma.
The Journal of experimental medicine
Takahashi T,Asano Y,Sugawara K,Yamashita T,Nakamura K,Saigusa R,Ichimura Y,Toyama T,Taniguchi T,Akamata K,Noda S,Yoshizaki A,Tsuruta D,Trojanowska M,Sato S
MA514127 was used in western blot to explore the contribution of epithelial cells to systemic sclerosis pathogenesis
Mon Apr 03 00:00:00 EDT 2017
HIV nucleoside reverse transcriptase inhibitors efavirenz and tenofovir change the growth and differentiation of primary gingival epithelium.
HIV medicine
Mitchell D,Israr M,Alam S,Dinello D,Kishel J,Jia R,Meyers C
MA5-14127 was used in immunohistochemistry to use organotypic gingival keratinocyte culture to explore the effects of HIV nucleoside reverse transcriptase inhibitors on the gingival epithelium
Tue Apr 01 00:00:00 EDT 2014
Keratin 79 identifies a novel population of migratory epithelial cells that initiates hair canal morphogenesis and regeneration.
Development (Cambridge, England)
Veniaminova NA,Vagnozzi AN,Kopinke D,Do TT,Murtaugh LC,Maillard I,Dlugosz AA,Reiter JF,Wong SY
MA5-14127 was used in immunohistochemistry to study the role of novel keratin 79-positive migratory epithelial cells in hair canal morphogenesis and regeneration
Sun Dec 01 00:00:00 EST 2013
Toward a molecular pathologic classification of urothelial carcinoma.
The American journal of pathology
Sjödahl G,Lövgren K,Lauss M,Patschan O,Gudjonsson S,Chebil G,Aine M,Eriksson P,Månsson W,Lindgren D,Fernö M,Liedberg F,Höglund M
MA5-14127 was used in immunohistochemistry (paraffin) to describe the molecular pathologic characteristics of urothelial carcinomas subtypes.
Sun Sep 01 00:00:00 EDT 2013
Keratin gene expression profiles after digit amputation in C57BL/6 vs. regenerative MRL mice imply an early regenerative keratinocyte activated-like state.
Physiological genomics
Cheng CH,Leferovich J,Zhang XM,Bedelbaeva K,Gourevitch D,Hatcher CJ,Basson CT,Heber-Katz E,Marx KA
MA5-14127 was used in immunohistochemistry to study the expression pattern of keratins in C57BL/6 mice and MRL regenerative mice before and after amputation of the middle digit
Mon Jun 03 00:00:00 EDT 2013
Epidermal growth factor-induced modulation of cytokeratin expression levels influences the morphological phenotype of head and neck squamous cell carcinoma cells.
Cell and tissue research
Makarova G,Bette M,Schmidt A,Jacob R,Cai C,Rodepeter F,Betz T,Sitterberg J,Bakowsky U,Moll R,Neff A,Sesterhenn A,Teymoortash A,Ocker M,Werner JA,Mandic R
MA5-14127 was used in western blot to study the effect on the morphology of head and neck squamous cell carcinoma cells of EGF-mediated regulation of cytokeratin expression
Tue Jan 01 00:00:00 EST 2013
Bioengineered skin humanized model of psoriasis.
Methods in molecular biology (Clifton, N.J.)
Carretero M,Guerrero-Aspizua S,Del Río M
MA5-14127 was used in immunohistochemistry to use bioengineering techniques to develop a murine skin humanized psoriasis model
Mon Jun 24 00:00:00 EDT 2013
Effect of the HIV nucleoside reverse transcriptase inhibitor zidovudine on the growth and differentiation of primary gingival epithelium.
HIV medicine
Mitchell D,Israr M,Alam S,Kishel J,Dinello D,Meyers C
MA5-14127 was used in immunohistochemistry to investigate the influence of zidovudine on gingival epithelium growth
Tue May 01 00:00:00 EDT 2012
Down-regulation of keratin 4 and keratin 13 expression in oral squamous cell carcinoma and epithelial dysplasia: a clue for histopathogenesis.
Histopathology
Sakamoto K,Aragaki T,Morita K,Kawachi H,Kayamori K,Nakanishi S,Omura K,Miki Y,Okada N,Katsube K,Takizawa T,Yamaguchi A
MA5-14127 was used in immunohistochemistry to study the expression of keratin 4 and keratin 13 in oral squamous cell carcinoma and epithelial dysplasia
Tue Mar 01 00:00:00 EST 2011
Development of skin-humanized mouse models of pachyonychia congenita.
The Journal of investigative dermatology
García M,Larcher F,Hickerson RP,Baselga E,Leachman SA,Kaspar RL,Del Rio M
MA5-14127 was used in immunohistochemistry to develop and evaluate a mouse model for pachyonychia congenita
Sun May 01 00:00:00 EDT 2011
The HIV protease inhibitor lopinavir/ritonavir (Kaletra) alters the growth, differentiation and proliferation of primary gingival epithelium.
HIV medicine
Israr M,Mitchell D,Alam S,Dinello D,Kishel JJ,Meyers C
MA5-14127 was used in immunohistochemistry to study the effects of an HIV protease inhibitor on the gingival epithelium.
Tue Mar 01 00:00:00 EST 2011
Multiple functions of Snail family genes during palate development in mice.
Development (Cambridge, England)
Murray SA,Oram KF,Gridley T
MA5-14127 was used in immunohistochemistry to study the multiple functions of Snail family genes during mouse palate development
Tue May 01 00:00:00 EDT 2007
New skin-equivalent model from de-epithelialized amnion membrane.
Cell and tissue research
Yang L,Shirakata Y,Shudou M,Dai X,Tokumaru S,Hirakawa S,Sayama K,Hamuro J,Hashimoto K
MA5-14127 was used in immunohistochemistry to develop a living skin-equivalent from de-epithelialized amnion membrane
Sun Oct 01 00:00:00 EDT 2006
Characterization of medroxyprogesterone and DMBA-induced multilineage mammary tumors by gene expression profiling.
Molecular carcinogenesis
Yin Y,Bai R,Russell RG,Beildeck ME,Xie Z,Kopelovich L,Glazer RI
MA5-14127 was used in immunohistochemistry to study the gene expression profile of medroxyprogesterone- and DMBA-induced multilineage mammary tumors
Thu Sep 01 00:00:00 EDT 2005
Abnormal development of thymic dendritic and epithelial cells in human X-linked severe combined immunodeficiency.
Clinical immunology (Orlando, Fla.)
Hale LP,Buckley RH,Puck JM,Patel DD
MA5-14127 was used in immunohistochemistry to study human X-linked severe combined immunodeficiency and thymic dendritic and epithelial cell development
Thu Jan 01 00:00:00 EST 2004
Corticosteroids regulate epithelial cell differentiation and Hassall body formation in the human thymus.
Journal of immunology (Baltimore, Md. : 1950)
Hale LP,Markert ML
MA5-14127 was used in immunohistochemistry to study the regulation of epithelial cell differentiation and human thymic medulla Hassall body formation by corticosteroids
Thu Jan 01 00:00:00 EST 2004
A study of cytokeratin profiles in localized cutaneous amyloids.
Archives of dermatological research
Chang YT,Liu HN,Wang WJ,Lee DD,Tsai SF
MA5-14127 was used in immunohistochemistry to investigate the expression profile of cytokeratins in cutaneous amyloids
Thu Jul 01 00:00:00 EDT 2004
Ku70/Ku80 and DNA-dependent protein kinase catalytic subunit modulate RAG-mediated cleavage: implications for the enforcement of the 12/23 rule.
The Journal of biological chemistry
Sawchuk DJ,Mansilla-Soto J,Alarcon C,Singha NC,Langen H,Bianchi ME,Lees-Miller SP,Nussenzweig MC,Cortes P
MA5-14127 was used in blocking or activating experiment to study the role of Ku70/80 and DNA-PK in regulating RAG-mediated cleavage and the significance for the 12/23 rule specificity
Fri Jul 09 00:00:00 EDT 2004