Invitrogen
MA1-91422 detects Cytomegalovirus from mouse samples.
MA1-91422 has been successfully used in immunohistochemistry (paraffin tissue) applications.
The MA1-91422 immunogen is: BALB/C mice were immunized with cytomegalovirus infected cell lysate.
Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).