Invitrogen
PA1-27382 detects Cytomegalovirus from virus samples. PA1-27382 will not react with HSV and VZV by indirect immunofluorescence.
PA1-27382 has been successfully used in immunofluorescence, and Western blot procedures.
The PA1-27382 immunogen is purified virions of strain AD 169.
Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5). Glycoprotein B (gB) is the main antigenic protein within the envelope of HCMV. It is required for HCMV mediated membrane fusion along with the gH/gL complex. The unprocessed precursor of HCMV gB is 906 amino acids in length, with a MW of approximately 100kDa.