Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, chicken and rat based on sequence homology.
This gene is located within a chromosomal region that shows loss of heterozygosity in some bladder cancers. It contains a 5' CpG island that may be a frequent target of hypermethylation, and it may undergo hypermethylation-based silencing in some bladder cancers.