Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 9
This antibody is predicted to react with bovine and porcine based on sequence homology.
Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bone proteoglycan II; Decorin; dermatan sulphate proteoglycans II; Dermatan sulphate proteoglycans II (DSPG2); PG-S2; PG40; proteoglycan core protein; Small leucine rich protein 1B; small leucine-rich protein 1B
Gene Aliases: CSCD; DC; DCN; DSPG2; mDcn; PG40; PGII; PGS2; SLRR1B
UniProt ID: (Human) P07585, (Mouse) P28654
Entrez Gene ID: (Human) 1634, (Mouse) 13179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support