Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MADAAPQLGK RKRELDVEEA HAASTEEKEA GVGNGTCAPV RLPFSGFRLQ KVLRESARDK IIFLHGKVNE ASEDGDGEDA VVILEKTPFQ VEQVAQLLTG SPELQLQFSN DIYSTYHLFP PRQLNDVKTT VVYPATEKHL QKYLRQDLRL IRETGDDYRN ITLPHLESQS LSIQWVYNIL DKKAEADRIV FENPDPSDGF VLIPDLKWNQ QQLDDLYLIA ICHRRGIRSL RDLTPEHLPL LRNILHQGQE AILQRYRMKG DHLRVYLHYL PSYYHLHVHF TALGFEAPGS GVERAHLLAE VIENLECDPR HYQQRTLTFA LRADDPLLKL LQEAQQS
Necessary for the complete degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. Removes the 7-methyl guanine cap structure from mRNA fragments shorter than 10 nucleotides that are produced by 3'->5' exosome-mediated mRNA decay. Releases m7GMP. Can also degrade m7GDP to m7GMP. Has no activity towards mRNA molecules longer than 25 nucleotides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-(N(7)-methyl 5'-triphosphoguanosine)-[mRNA] diphosphatase; DCS-1; Decapping scavenger enzyme; heat shock-like protein 1; HINT-5; Hint-related 7meGMP-directed hydrolase; Histidine triad nucleotide-binding protein 5; Histidine triad protein member 5; homolog of C. elegans 7meGMP-directed hydrolase dcs-1; m7GpppX diphosphatase; mRNA decapping enzyme; Scavenger mRNA-decapping enzyme DcpS
Gene Aliases: ARS; DCPS; DCS1; HINT-5; HINT5; HSL1; HSPC015
UniProt ID: (Human) Q96C86, (Rat) Q8K4F7
Entrez Gene ID: (Human) 28960, (Rat) 266605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support