Based on 100% sequence identity, this antibody is predicted to react with Orangutan
METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.