Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee
HNF1A is a transcription factor required for the expression of several liver-specific genes. This protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'.