Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: AFRICAFNAQ RLTLAKVARE QVMDTLVRIL ARCDIMVLQE VVDSSGSAIP LLLRELNRFD GSGPYSTLSS PQLGRSTYME TYVYFYRSHK TQVLSSYVYN DEDDVFAREP FVAQFSLPSN VLPSLVLVPL HTTPKAVEKE LNALYDVFLE VSQHWQSKDV ILLGDFNADC ASLTKKRLDK LELRTEPGFH WVIADGEDTT VRASTHCTYD RVVLHGERCR SLLHTAAAFD FPTSFQLTEE EALNISDHYP VEVELKLSQA HSVQPLSLTV LLLLSLLSPQ LCPAA
DNASE1L1 encodes a deoxyribonuclease protein that shows high sequence similarity to DNase I. The encoded protein is localized to the endoplasmic reticulum and modified by N-linked glycosylation. Alternate transcriptional splice variants encoding the same protein have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Deoxyribonuclease I-like 1; Deoxyribonuclease-1-like 1; DNase I, lysosomal-like; DNase I-like 1; DNase I-like, muscle-specific; DNase X; Muscle-specific DNase I-like; XIB
Gene Aliases: DNAS1L1; DNASE1L1; DNASEX; DNL1L; G4.8; XIB
UniProt ID: (Human) P49184
Entrez Gene ID: (Human) 1774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support