Search Thermo Fisher Scientific
61003S is a mixture of monoclonal antibodies that target Desmoplakin 1/2.
Tumors specifically detected: Primary and metastatic carcinoma and meningioma. Polypeptides reacting: Desmoplakin 1 and 2 (MW 250 000 and 215 000).
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.