Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute lyophilized product with 1 mL distilled water.
DP2.17 shows distinct punctate membrane staining of epithelia. DP2.17 reacts with the rod fragment of desmoplakin 1. Polypeptide reacting: Desmoplakin no. 1 (MW 250,000 polypeptide of desmosomal plaques) Reactivity on cultured cell lines: Several human carcinoma cell lines, such as MCF-7, A-431, TR-146; bovine cells: MDBK, BMGE Tumors specifically detected: Primary and metastatic carcinoma; meningioma.
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 250/210 kDa paraneoplastic pemphigus antigen; Desmoplakin; desmoplakin I/II; Desmoplakin II; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP
Gene Aliases: 2300002E22Rik; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; DP; DSP; rul
UniProt ID: (Human) P15924, (Mouse) E9Q557
Entrez Gene ID: (Human) 1832, (Bovine) 514360, (Chicken) 420869, (Mouse) 109620, (Rat) 306871
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support