Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
0.5-2 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Mouse |
Host/Isotype |
Mouse / IgG2a |
Class |
Recombinant Monoclonal |
Type |
Antibody |
Immunogen |
This antibody was raised against amino acids 1638-1899 of mouse Dicer (aka dicer 1, ribonuclease type III). |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
1 mg/mL |
Purification |
Protein A |
Storage buffer |
140mM HEPES/32mM sodium acetate, pH 7.5, with 30% glycerol, 70mM NaCl |
Contains |
0.035% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_2793815 |
Dicer is a double-stranded RNA (dsRNA) endoribonuclease playing a central role in short dsRNA-mediated post-transcriptional gene silencing. It cleaves naturally occurring long dsRNAs and short hairpin pre-microRNAs (miRNA) into fragments of twenty-one to twenty-three nucleotides with 3' overhang of two nucleotides, producing respectively short interfering RNAs (siRNA) and mature microRNAs. SiRNAs and miRNAs serve as guide to direct the RNA-induced silencing complex (RISC) to complementary RNAs to degrade them or prevent their translation. Gene silencing mediated by siRNAs, also called RNA interference, controls the elimination of transcripts from mobile and repetitive DNA elements of the genome but also the degradation of exogenous RNA of viral origin for instance. The miRNA pathway on the other side is a means to specifically regulate the expression of target genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DCR; DCR-1; Dicer1, Dcr-1 homolog; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; helicase-moi
Gene Aliases: 1110006F08Rik; D12Ertd7e; mKIAA0928
Entrez Gene ID: (Mouse) 192119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support