Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa, rat brain cytosol.
This antibody is expected to cross-react with mouse (100%) and chicken (85%) due to sequence homology.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a cytoplasmic actin-binding protein thought to play a role in the process of neuronal growth. It is a member of the drebrin family of proteins that are developmentally regulated in the brain. A decrease in the amount of this protein in the brain has been implicated as a possible contributing factor in the pathogenesis of memory disturbance in Alzheimer's disease. At least two alternative splice variants encoding different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Developmentally-regulated brain protein; DKFZp434D064; Drebrin; drebrin E; drebrin E2; drebrin-1
Gene Aliases: D0S117E; DBN1
UniProt ID: (Human) Q16643, (Rat) Q07266
Entrez Gene ID: (Human) 1627, (Rat) 81653
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support