Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
The protein encoded by this gene is a preproprotein that is proteolytically processed to form the secreted opioid peptides beta-neoendorphin, dynorphin, leu-enkephalin, rimorphin, and leumorphin. These peptides are ligands for the kappa-type of opioid receptor. Dynorphin is involved in modulating responses to several psychoactive substances, including cocaine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; big dynorphin; dynorphin A(1-13); dynorphin A(1-17); dynorphin A(1-8); leu-enkephalin; leumorphin; MGC26418; neoendorphin-dynorphin-enkephalin prepropeptide; Preprodynorphin; preproenkephalin B; proenkephalin B; Proenkephalin-B; rimorphin
Gene Aliases: ADCA; Dyn; PDYN; PENKB; SCA23
UniProt ID: (Human) P01213
Entrez Gene ID: (Human) 5173, (Mouse) 18610, (Rat) 29190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support